Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 2828518-2828785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUM3
Entrez Gene ID
9933
Gene Name
pumilio RNA binding family member 3
Gene Aliases
HA-8, HLA-HA8, KIAA0020, PEN, PUF-A, PUF6, XTP5
UniGene
Hs.493309
Gene Chromosome Location
Chr.9: 2804152-2844130 on Build GRCh38
Transcripts
NM_014878.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4152239 c.946A>G p.M316V Chr.9: 2828685-2828685 on Build GRCh38
4419665 c.889G>T p.V297L Chr.9: 2828742-2828742 on Build GRCh38
6260357 c.931C>A p.Q311K Chr.9: 2828700-2828700 on Build GRCh38
4561470 c.886G>A p.E296K Chr.9: 2828745-2828745 on Build GRCh38
2771859 c.888G>A p.E296E Chr.9: 2828743-2828743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539912169 Chr.9: 2828557-2828557 on Build GRCh38
rs200312927 Chr.9: 2828686-2828686 on Build GRCh38
rs763818690 Chr.9: 2828743-2828743 on Build GRCh38
rs555602821 Chr.9: 2828716-2828716 on Build GRCh38
rs754814072 Chr.9: 2828766-2828766 on Build GRCh38
rs761401968 Chr.9: 2828746-2828746 on Build GRCh38
rs370781388 Chr.9: 2828647-2828647 on Build GRCh38
rs184436153 Chr.9: 2828641-2828641 on Build GRCh38
rs978580898 Chr.9: 2828594-2828594 on Build GRCh38
rs188123994 Chr.9: 2828690-2828690 on Build GRCh38
rs966746590 Chr.9: 2828567-2828567 on Build GRCh38
rs773457579 Chr.9: 2828563-2828563 on Build GRCh38
rs766993196 Chr.9: 2828755-2828755 on Build GRCh38
rs763658753 Chr.9: 2828652-2828652 on Build GRCh38
rs2173904 Chr.9: 2828765-2828765 on Build GRCh38
rs386731748 Chr.9: 2828765-2828767 on Build GRCh38
rs746378439 Chr.9: 2828734-2828734 on Build GRCh38
rs915095863 Chr.9: 2828560-2828560 on Build GRCh38
rs7849647 Chr.9: 2828548-2828548 on Build GRCh38
rs137936878 Chr.9: 2828726-2828726 on Build GRCh38
rs376706931 Chr.9: 2828685-2828685 on Build GRCh38
rs537493626 Chr.9: 2828738-2828738 on Build GRCh38
rs2270891 Chr.9: 2828742-2828742 on Build GRCh38
rs758646587 Chr.9: 2828717-2828717 on Build GRCh38
rs539265165 Chr.9: 2828556-2828556 on Build GRCh38
rs936656305 Chr.9: 2828623-2828623 on Build GRCh38
rs777778720 Chr.9: 2828718-2828718 on Build GRCh38
rs372399592 Chr.9: 2828664-2828664 on Build GRCh38
rs756005709 Chr.9: 2828762-2828762 on Build GRCh38
rs985755420 Chr.9: 2828687-2828687 on Build GRCh38
rs777185887 Chr.9: 2828732-2828732 on Build GRCh38
rs944218481 Chr.9: 2828700-2828700 on Build GRCh38
rs761127896 Chr.9: 2828625-2828625 on Build GRCh38
rs777047873 Chr.9: 2828636-2828636 on Build GRCh38
rs755923388 Chr.9: 2828714-2828714 on Build GRCh38
rs906401912 Chr.9: 2828656-2828656 on Build GRCh38
rs143778583 Chr.9: 2828694-2828694 on Build GRCh38
rs201465798 Chr.9: 2828745-2828745 on Build GRCh38
rs540277318 Chr.9: 2828757-2828757 on Build GRCh38
rs771447261 Chr.9: 2828632-2828632 on Build GRCh38
rs759944730 Chr.9: 2828639-2828639 on Build GRCh38
rs925342240 Chr.9: 2828604-2828604 on Build GRCh38
rs147348344 Chr.9: 2828715-2828715 on Build GRCh38

View Full Product Details