Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42631138-42631363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928323
Entrez Gene ID
101928323
Gene Name
uncharacterized LOC101928323
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.3: 42612310-42654388 on Build GRCh38
Transcripts
NR_125400.1 NM_005385.3 XM_005265173.1 XM_006713171.1 XM_006713173.2 XM_011533747.2 XM_011533750.2 XM_017006474.1 XM_017006475.1 XM_017006476.1 XM_017006477.1 XM_017006478.1 XM_017006479.1 XM_017006480.1

Gene Symbol
NKTR
Entrez Gene ID
4820
Gene Name
natural killer cell triggering receptor
Gene Aliases
p104
UniGene
Hs.529509
Gene Chromosome Location
Chr.3: 42600655-42648741 on Build GRCh38
Transcripts
NR_125400.1 NM_005385.3 XM_005265173.1 XM_006713171.1 XM_006713173.2 XM_011533747.2 XM_011533750.2 XM_017006474.1 XM_017006475.1 XM_017006476.1 XM_017006477.1 XM_017006478.1 XM_017006479.1 XM_017006480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229832 c.505C>T p.R169* Chr.3: 42631271-42631271 on Build GRCh38
5864529 c.481G>A p.A161T Chr.3: 42631247-42631247 on Build GRCh38
6223249 c.487A>C p.R163R Chr.3: 42631253-42631253 on Build GRCh38
1044712 c.512T>C p.I171T Chr.3: 42631278-42631278 on Build GRCh38
1580063 c.536A>C p.K179T Chr.3: 42631302-42631302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764791700 Chr.3: 42631264-42631264 on Build GRCh38
rs764568126 Chr.3: 42631217-42631217 on Build GRCh38
rs369564869 Chr.3: 42631336-42631336 on Build GRCh38
rs750113249 Chr.3: 42631278-42631278 on Build GRCh38
rs528046789 Chr.3: 42631307-42631307 on Build GRCh38
rs779751604 Chr.3: 42631311-42631311 on Build GRCh38
rs754776862 Chr.3: 42631335-42631335 on Build GRCh38
rs751366336 Chr.3: 42631323-42631323 on Build GRCh38
rs761097980 Chr.3: 42631210-42631210 on Build GRCh38
rs781145072 Chr.3: 42631337-42631337 on Build GRCh38
rs201863592 Chr.3: 42631240-42631240 on Build GRCh38
rs767941538 Chr.3: 42631177-42631177 on Build GRCh38
rs889379755 Chr.3: 42631297-42631297 on Build GRCh38
rs376753039 Chr.3: 42631220-42631220 on Build GRCh38
rs1040599060 Chr.3: 42631284-42631284 on Build GRCh38
rs910767006 Chr.3: 42631241-42631241 on Build GRCh38
rs775652772 Chr.3: 42631196-42631196 on Build GRCh38
rs781019320 Chr.3: 42631334-42631335 on Build GRCh38
rs943524170 Chr.3: 42631272-42631272 on Build GRCh38

View Full Product Details