Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116918024-116918249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX6
Entrez Gene ID
222546
Gene Name
regulatory factor X6
Gene Aliases
MTCHRS, MTFS, RFXDC1, dJ955L16.1
UniGene
Hs.352276
Gene Chromosome Location
Chr.6: 116877213-116932163 on Build GRCh38
Transcripts
NM_173560.3 XM_011535589.1 XM_017010477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4185581 c.985G>A p.V329I Chr.6: 116918049-116918049 on Build GRCh38
6172053 c.997G>T p.A333S Chr.6: 116918061-116918061 on Build GRCh38
7251690 c.1022+5G>T p.? Chr.6: 116918091-116918091 on Build GRCh38
5413892 c.994C>T p.P332S Chr.6: 116918058-116918058 on Build GRCh38
4616921 c.1014G>T p.M338I Chr.6: 116918078-116918078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746808009 Chr.6: 116918121-116918121 on Build GRCh38
rs965878014 Chr.6: 116918152-116918152 on Build GRCh38
rs749913626 Chr.6: 116918107-116918107 on Build GRCh38
rs758437646 Chr.6: 116918111-116918111 on Build GRCh38
rs780088838 Chr.6: 116918120-116918120 on Build GRCh38
rs753499358 Chr.6: 116918078-116918078 on Build GRCh38
rs752117103 Chr.6: 116918118-116918120 on Build GRCh38
rs189126346 Chr.6: 116918206-116918206 on Build GRCh38
rs919608834 Chr.6: 116918166-116918166 on Build GRCh38
rs980426503 Chr.6: 116918146-116918146 on Build GRCh38
rs538580820 Chr.6: 116918177-116918177 on Build GRCh38
rs756920384 Chr.6: 116918079-116918079 on Build GRCh38
rs184373346 Chr.6: 116918105-116918105 on Build GRCh38

View Full Product Details