Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129137531-129137756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPA
Entrez Gene ID
5524
Gene Name
protein phosphatase 2 phosphatase activator
Gene Aliases
PP2A, PPP2R4, PR53
UniGene
Hs.400740
Gene Chromosome Location
Chr.9: 129110949-129148946 on Build GRCh38
Transcripts
NM_001193397.1 NM_001271832.1 NM_021131.4 NM_178000.2 NM_178001.2 NM_178003.2 XM_011518834.2 XM_011518835.2 XM_011518836.2 XM_011518837.2 XM_011518838.2 XM_017014888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6878607 c.833C>T p.A278V Chr.9: 129137634-129137634 on Build GRCh38
5410514 c.867G>A p.L289L Chr.9: 129137668-129137668 on Build GRCh38
3654814 c.888C>T p.T296T Chr.9: 129137689-129137689 on Build GRCh38
4965219 c.823G>A p.D275N Chr.9: 129137624-129137624 on Build GRCh38
1460394 c.834C>T p.A278A Chr.9: 129137635-129137635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934729699 Chr.9: 129137631-129137631 on Build GRCh38
rs200171889 Chr.9: 129137656-129137656 on Build GRCh38
rs985521781 Chr.9: 129137581-129137581 on Build GRCh38
rs201904712 Chr.9: 129137641-129137641 on Build GRCh38
rs756302314 Chr.9: 129137614-129137614 on Build GRCh38
rs530974957 Chr.9: 129137671-129137671 on Build GRCh38
rs76263259 Chr.9: 129137549-129137549 on Build GRCh38
rs757931542 Chr.9: 129137689-129137689 on Build GRCh38
rs771545360 Chr.9: 129137560-129137560 on Build GRCh38
rs957729138 Chr.9: 129137579-129137579 on Build GRCh38
rs150647017 Chr.9: 129137635-129137635 on Build GRCh38
rs548664652 Chr.9: 129137620-129137620 on Build GRCh38
rs575122036 Chr.9: 129137568-129137568 on Build GRCh38
rs778674089 Chr.9: 129137660-129137660 on Build GRCh38
rs984209938 Chr.9: 129137663-129137663 on Build GRCh38
rs895795218 Chr.9: 129137735-129137735 on Build GRCh38
rs776425459 Chr.9: 129137582-129137582 on Build GRCh38
rs924650382 Chr.9: 129137599-129137599 on Build GRCh38
rs770925567 Chr.9: 129137704-129137704 on Build GRCh38
rs937409559 Chr.9: 129137695-129137695 on Build GRCh38
rs550932260 Chr.9: 129137729-129137729 on Build GRCh38
rs767343284 Chr.9: 129137561-129137561 on Build GRCh38
rs140781354 Chr.9: 129137659-129137659 on Build GRCh38
rs3118631 Chr.9: 129137567-129137567 on Build GRCh38
rs747720753 Chr.9: 129137682-129137682 on Build GRCh38
rs763219015 Chr.9: 129137732-129137732 on Build GRCh38
rs767777862 Chr.9: 129137644-129137644 on Build GRCh38
rs769446642 Chr.9: 129137720-129137720 on Build GRCh38
rs759248164 Chr.9: 129137583-129137583 on Build GRCh38
rs965677095 Chr.9: 129137611-129137611 on Build GRCh38
rs746957818 Chr.9: 129137698-129137698 on Build GRCh38
rs370634207 Chr.9: 129137608-129137608 on Build GRCh38
rs763166988 Chr.9: 129137616-129137616 on Build GRCh38
rs376576459 Chr.9: 129137587-129137587 on Build GRCh38
rs776422724 Chr.9: 129137712-129137712 on Build GRCh38
rs770729655 Chr.9: 129137569-129137569 on Build GRCh38
rs745727199 Chr.9: 129137719-129137719 on Build GRCh38

View Full Product Details