Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45083168-45083429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACAD
Entrez Gene ID
23148
Gene Name
NAC alpha domain containing
Gene Aliases
-
UniGene
Hs.96633
Gene Chromosome Location
Chr.7: 45080432-45089443 on Build GRCh38
Transcripts
NM_001146334.1 XM_006715674.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276862 c.2859C>T p.G953G Chr.7: 45083321-45083321 on Build GRCh38
7233678 c.2792C>G p.T931R Chr.7: 45083388-45083388 on Build GRCh38
4646797 c.3012G>A p.Q1004Q Chr.7: 45083168-45083168 on Build GRCh38
3832878 c.2795G>A p.G932D Chr.7: 45083385-45083385 on Build GRCh38
6378151 c.2919A>C p.L973F Chr.7: 45083261-45083261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032317099 Chr.7: 45083240-45083240 on Build GRCh38
rs1001377898 Chr.7: 45083216-45083216 on Build GRCh38
rs770278908 Chr.7: 45083224-45083224 on Build GRCh38
rs776350027 Chr.7: 45083285-45083285 on Build GRCh38
rs765177591 Chr.7: 45083312-45083312 on Build GRCh38
rs760363390 Chr.7: 45083260-45083260 on Build GRCh38
rs771508797 Chr.7: 45083254-45083254 on Build GRCh38
rs750349275 Chr.7: 45083373-45083373 on Build GRCh38
rs932728151 Chr.7: 45083192-45083192 on Build GRCh38
rs529749369 Chr.7: 45083210-45083210 on Build GRCh38
rs534777748 Chr.7: 45083401-45083401 on Build GRCh38
rs975909425 Chr.7: 45083325-45083325 on Build GRCh38
rs1010800394 Chr.7: 45083297-45083297 on Build GRCh38
rs773596992 Chr.7: 45083231-45083231 on Build GRCh38
rs985993117 Chr.7: 45083289-45083289 on Build GRCh38
rs570748050 Chr.7: 45083381-45083381 on Build GRCh38
rs904890222 Chr.7: 45083309-45083309 on Build GRCh38
rs919089513 Chr.7: 45083364-45083364 on Build GRCh38
rs548119377 Chr.7: 45083211-45083211 on Build GRCh38
rs1052273661 Chr.7: 45083232-45083234 on Build GRCh38
rs558988221 Chr.7: 45083205-45083205 on Build GRCh38
rs139828162 Chr.7: 45083282-45083282 on Build GRCh38
rs563180444 Chr.7: 45083272-45083272 on Build GRCh38
rs775105208 Chr.7: 45083259-45083259 on Build GRCh38
rs751829108 Chr.7: 45083388-45083388 on Build GRCh38
rs745347677 Chr.7: 45083253-45083253 on Build GRCh38
rs758481325 Chr.7: 45083376-45083376 on Build GRCh38
rs902555680 Chr.7: 45083209-45083209 on Build GRCh38
rs143697186 Chr.7: 45083299-45083299 on Build GRCh38

View Full Product Details