Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122698103-122698358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf120
Entrez Gene ID
399814
Gene Name
chromosome 10 open reading frame 120
Gene Aliases
bA318C4.1
UniGene
Hs.363649
Gene Chromosome Location
Chr.10: 122697709-122699822 on Build GRCh38
Transcripts
NM_001010912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6065394 c.425T>A p.I142K Chr.10: 122698316-122698316 on Build GRCh38
229177 c.533G>A p.G178E Chr.10: 122698208-122698208 on Build GRCh38
4553575 c.594G>A p.V198V Chr.10: 122698147-122698147 on Build GRCh38
915711 c.544C>T p.P182S Chr.10: 122698197-122698197 on Build GRCh38
4012345 c.543G>T p.Q181H Chr.10: 122698198-122698198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778602192 Chr.10: 122698151-122698151 on Build GRCh38
rs750474765 Chr.10: 122698240-122698240 on Build GRCh38
rs754645575 Chr.10: 122698294-122698294 on Build GRCh38
rs545427301 Chr.10: 122698298-122698298 on Build GRCh38
rs758266643 Chr.10: 122698314-122698314 on Build GRCh38
rs546338112 Chr.10: 122698252-122698252 on Build GRCh38
rs1051469629 Chr.10: 122698177-122698177 on Build GRCh38
rs763911756 Chr.10: 122698319-122698319 on Build GRCh38
rs751310217 Chr.10: 122698329-122698329 on Build GRCh38
rs569498958 Chr.10: 122698317-122698317 on Build GRCh38
rs753731837 Chr.10: 122698142-122698142 on Build GRCh38
rs61729644 Chr.10: 122698251-122698251 on Build GRCh38
rs577934787 Chr.10: 122698218-122698218 on Build GRCh38
rs761518777 Chr.10: 122698163-122698165 on Build GRCh38
rs200087733 Chr.10: 122698233-122698233 on Build GRCh38
rs144052210 Chr.10: 122698170-122698170 on Build GRCh38
rs139344663 Chr.10: 122698293-122698293 on Build GRCh38
rs141554298 Chr.10: 122698217-122698217 on Build GRCh38
rs560814531 Chr.10: 122698229-122698229 on Build GRCh38
rs766770680 Chr.10: 122698232-122698235 on Build GRCh38
rs201845744 Chr.10: 122698225-122698225 on Build GRCh38
rs138322078 Chr.10: 122698134-122698134 on Build GRCh38
rs757475654 Chr.10: 122698140-122698140 on Build GRCh38
rs770441464 Chr.10: 122698207-122698207 on Build GRCh38
rs376780812 Chr.10: 122698214-122698214 on Build GRCh38
rs531792820 Chr.10: 122698295-122698295 on Build GRCh38
rs747926166 Chr.10: 122698169-122698169 on Build GRCh38
rs911487442 Chr.10: 122698310-122698310 on Build GRCh38
rs144060316 Chr.10: 122698305-122698305 on Build GRCh38
rs776957683 Chr.10: 122698236-122698237 on Build GRCh38
rs759844183 Chr.10: 122698266-122698268 on Build GRCh38
rs148798284 Chr.10: 122698209-122698209 on Build GRCh38
rs370374331 Chr.10: 122698215-122698215 on Build GRCh38
rs749796267 Chr.10: 122698285-122698285 on Build GRCh38
rs765267363 Chr.10: 122698245-122698245 on Build GRCh38
rs754320545 Chr.10: 122698272-122698272 on Build GRCh38
rs563017827 Chr.10: 122698291-122698291 on Build GRCh38
rs756897728 Chr.10: 122698265-122698265 on Build GRCh38
rs781314088 Chr.10: 122698141-122698141 on Build GRCh38
rs765763482 Chr.10: 122698316-122698317 on Build GRCh38
rs374030682 Chr.10: 122698234-122698234 on Build GRCh38
rs777486593 Chr.10: 122698264-122698264 on Build GRCh38
rs371446380 Chr.10: 122698283-122698283 on Build GRCh38
rs202094699 Chr.10: 122698266-122698266 on Build GRCh38
rs745435403 Chr.10: 122698271-122698271 on Build GRCh38
rs776144555 Chr.10: 122698208-122698208 on Build GRCh38
rs374585431 Chr.10: 122698284-122698284 on Build GRCh38
rs764456744 Chr.10: 122698128-122698128 on Build GRCh38
rs754892357 Chr.10: 122698241-122698241 on Build GRCh38
rs890139625 Chr.10: 122698179-122698179 on Build GRCh38
rs746651873 Chr.10: 122698191-122698191 on Build GRCh38
rs368917352 Chr.10: 122698308-122698308 on Build GRCh38
rs147336593 Chr.10: 122698187-122698187 on Build GRCh38
rs754876866 Chr.10: 122698143-122698143 on Build GRCh38
rs773929863 Chr.10: 122698149-122698149 on Build GRCh38
rs567759719 Chr.10: 122698201-122698201 on Build GRCh38

View Full Product Details