Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118660702-118660961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TREH
Entrez Gene ID
11181
Gene Name
trehalase
Gene Aliases
TRE, TREA
UniGene
Hs.129712
Gene Chromosome Location
Chr.11: 118658232-118679672 on Build GRCh38
Transcripts
NM_001301065.1 NM_007180.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585433 c.546G>A p.R182R Chr.11: 118660726-118660726 on Build GRCh38
923643 c.915G>A p.R305R Chr.11: 118660726-118660726 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782171768 Chr.11: 118660754-118660754 on Build GRCh38
rs782261696 Chr.11: 118660727-118660727 on Build GRCh38
rs782202854 Chr.11: 118660733-118660733 on Build GRCh38
rs782089073 Chr.11: 118660740-118660740 on Build GRCh38
rs782459761 Chr.11: 118660846-118660846 on Build GRCh38
rs782607384 Chr.11: 118660725-118660725 on Build GRCh38
rs782246400 Chr.11: 118660912-118660912 on Build GRCh38
rs933231292 Chr.11: 118660765-118660765 on Build GRCh38
rs782438204 Chr.11: 118660916-118660916 on Build GRCh38
rs781883020 Chr.11: 118660924-118660925 on Build GRCh38
rs782023814 Chr.11: 118660729-118660729 on Build GRCh38
rs782304296 Chr.11: 118660927-118660927 on Build GRCh38
rs201279957 Chr.11: 118660728-118660728 on Build GRCh38
rs782429475 Chr.11: 118660782-118660782 on Build GRCh38
rs572792365 Chr.11: 118660767-118660767 on Build GRCh38
rs891158247 Chr.11: 118660775-118660775 on Build GRCh38
rs782233961 Chr.11: 118660877-118660877 on Build GRCh38
rs782012163 Chr.11: 118660919-118660919 on Build GRCh38
rs782338713 Chr.11: 118660882-118660882 on Build GRCh38
rs782792249 Chr.11: 118660766-118660766 on Build GRCh38
rs372828123 Chr.11: 118660821-118660821 on Build GRCh38
rs782314286 Chr.11: 118660737-118660737 on Build GRCh38
rs941873148 Chr.11: 118660721-118660721 on Build GRCh38
rs893234651 Chr.11: 118660838-118660838 on Build GRCh38
rs544343788 Chr.11: 118660935-118660935 on Build GRCh38
rs782009103 Chr.11: 118660901-118660901 on Build GRCh38
rs781992305 Chr.11: 118660752-118660752 on Build GRCh38
rs782776610 Chr.11: 118660749-118660749 on Build GRCh38
rs782573306 Chr.11: 118660869-118660869 on Build GRCh38
rs1020413982 Chr.11: 118660892-118660892 on Build GRCh38
rs782752988 Chr.11: 118660816-118660816 on Build GRCh38
rs782527651 Chr.11: 118660832-118660832 on Build GRCh38
rs373328415 Chr.11: 118660923-118660923 on Build GRCh38
rs781968277 Chr.11: 118660738-118660738 on Build GRCh38

View Full Product Details