Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 138560712-138560973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPOPL
Entrez Gene ID
339745
Gene Name
speckle type BTB/POZ protein like
Gene Aliases
BTBD33
UniGene
Hs.333297
Gene Chromosome Location
Chr.2: 138501780-138573235 on Build GRCh38
Transcripts
NM_001001664.2 XM_005263655.4 XM_005263656.4 XM_005263657.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3379779 c.776A>C p.Y259S Chr.2: 138560866-138560866 on Build GRCh38
6413256 c.826G>A p.A276T Chr.2: 138560916-138560916 on Build GRCh38
4392493 c.744T>C p.P248P Chr.2: 138560834-138560834 on Build GRCh38
6018991 c.715-8G>A p.? Chr.2: 138560797-138560797 on Build GRCh38
3117058 c.784A>G p.R262G Chr.2: 138560874-138560874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753384840 Chr.2: 138560795-138560795 on Build GRCh38
rs746021194 Chr.2: 138560763-138560765 on Build GRCh38
rs761501214 Chr.2: 138560938-138560938 on Build GRCh38
rs374888829 Chr.2: 138560770-138560770 on Build GRCh38
rs567835124 Chr.2: 138560918-138560918 on Build GRCh38
rs757149030 Chr.2: 138560747-138560747 on Build GRCh38
rs66551604 Chr.2: 138560745-138560745 on Build GRCh38
rs35560905 Chr.2: 138560763-138560763 on Build GRCh38
rs78240832 Chr.2: 138560774-138560775 on Build GRCh38
rs368360974 Chr.2: 138560785-138560785 on Build GRCh38
rs781153637 Chr.2: 138560790-138560791 on Build GRCh38
rs397868606 Chr.2: 138560775-138560775 on Build GRCh38
rs1041856585 Chr.2: 138560893-138560893 on Build GRCh38
rs749448528 Chr.2: 138560936-138560936 on Build GRCh38
rs774071344 Chr.2: 138560850-138560850 on Build GRCh38
rs767028198 Chr.2: 138560787-138560787 on Build GRCh38
rs201949512 Chr.2: 138560810-138560810 on Build GRCh38
rs768598451 Chr.2: 138560814-138560814 on Build GRCh38
rs778875957 Chr.2: 138560763-138560764 on Build GRCh38
rs899764272 Chr.2: 138560858-138560858 on Build GRCh38
rs781497682 Chr.2: 138560827-138560827 on Build GRCh38
rs772064165 Chr.2: 138560873-138560873 on Build GRCh38
rs759036296 Chr.2: 138560809-138560809 on Build GRCh38
rs551503739 Chr.2: 138560794-138560794 on Build GRCh38
rs770713424 Chr.2: 138560849-138560849 on Build GRCh38
rs778237376 Chr.2: 138560762-138560762 on Build GRCh38
rs775259509 Chr.2: 138560885-138560885 on Build GRCh38
rs151051467 Chr.2: 138560846-138560846 on Build GRCh38
rs780780491 Chr.2: 138560791-138560791 on Build GRCh38
rs745648849 Chr.2: 138560870-138560870 on Build GRCh38
rs374939865 Chr.2: 138560798-138560798 on Build GRCh38
rs760097331 Chr.2: 138560793-138560793 on Build GRCh38
rs763474280 Chr.2: 138560897-138560897 on Build GRCh38
rs750101442 Chr.2: 138560796-138560796 on Build GRCh38
rs757437982 Chr.2: 138560762-138560763 on Build GRCh38
rs564888975 Chr.2: 138560808-138560808 on Build GRCh38
rs760280026 Chr.2: 138560886-138560886 on Build GRCh38

View Full Product Details