Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10552040-10552295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4D
Entrez Gene ID
84971
Gene Name
autophagy related 4D cysteine peptidase
Gene Aliases
APG4-D, APG4D, AUTL4
UniGene
Hs.512799
Gene Chromosome Location
Chr.19: 10543894-10553423 on Build GRCh38
Transcripts
NM_001281504.1 NM_032885.5 NR_104024.1 NR_104025.1 XM_006722927.1 NR_031603.1

Gene Symbol
MIR1238
Entrez Gene ID
100302226
Gene Name
microRNA 1238
Gene Aliases
MIRN1238, hsa-mir-1238
UniGene
Gene Chromosome Location
Chr.19: 10552122-10552204 on Build GRCh38
Transcripts
NM_001281504.1 NM_032885.5 NR_104024.1 NR_104025.1 XM_006722927.1 NR_031603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606723 c.1122G>A p.E374E Chr.19: 10552121-10552121 on Build GRCh38
990619 c.1046-2A>G p.? Chr.19: 10552043-10552043 on Build GRCh38
4073883 c.1057C>T p.L353L Chr.19: 10552056-10552056 on Build GRCh38
7001598 c.1109_1110ins11 p.D370fs*47 Chr.19: 10552108-10552109 on Build GRCh38
3528292 c.1050C>T p.D350D Chr.19: 10552049-10552049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374913540 Chr.19: 10552188-10552188 on Build GRCh38
rs369002764 Chr.19: 10552219-10552219 on Build GRCh38
rs769601412 Chr.19: 10552174-10552174 on Build GRCh38
rs201291151 Chr.19: 10552265-10552265 on Build GRCh38
rs758790754 Chr.19: 10552218-10552218 on Build GRCh38
rs368969533 Chr.19: 10552146-10552146 on Build GRCh38
rs199912582 Chr.19: 10552159-10552159 on Build GRCh38
rs765558252 Chr.19: 10552160-10552160 on Build GRCh38
rs746490907 Chr.19: 10552154-10552154 on Build GRCh38
rs376707293 Chr.19: 10552189-10552189 on Build GRCh38
rs760450241 Chr.19: 10552192-10552192 on Build GRCh38
rs769534266 Chr.19: 10552106-10552106 on Build GRCh38
rs150808873 Chr.19: 10552226-10552226 on Build GRCh38
rs766736761 Chr.19: 10552221-10552221 on Build GRCh38
rs755064220 Chr.19: 10552167-10552167 on Build GRCh38
rs772780311 Chr.19: 10552118-10552118 on Build GRCh38
rs201531397 Chr.19: 10552168-10552168 on Build GRCh38
rs761538562 Chr.19: 10552104-10552104 on Build GRCh38
rs964300302 Chr.19: 10552078-10552078 on Build GRCh38
rs765893329 Chr.19: 10552132-10552132 on Build GRCh38
rs763826655 Chr.19: 10552196-10552196 on Build GRCh38
rs142513981 Chr.19: 10552264-10552264 on Build GRCh38
rs537782433 Chr.19: 10552183-10552183 on Build GRCh38
rs767215101 Chr.19: 10552154-10552154 on Build GRCh38
rs141346756 Chr.19: 10552121-10552121 on Build GRCh38
rs780638134 Chr.19: 10552163-10552163 on Build GRCh38
rs148200963 Chr.19: 10552103-10552103 on Build GRCh38
rs750709640 Chr.19: 10552211-10552211 on Build GRCh38
rs770777634 Chr.19: 10552248-10552248 on Build GRCh38
rs748970122 Chr.19: 10552247-10552247 on Build GRCh38
rs371715950 Chr.19: 10552158-10552158 on Build GRCh38
rs758778787 Chr.19: 10552162-10552162 on Build GRCh38
rs756243375 Chr.19: 10552233-10552233 on Build GRCh38
rs748434288 Chr.19: 10552171-10552171 on Build GRCh38
rs376871668 Chr.19: 10552270-10552270 on Build GRCh38
rs750917610 Chr.19: 10552161-10552161 on Build GRCh38
rs749131513 Chr.19: 10552184-10552184 on Build GRCh38
rs747111031 Chr.19: 10552092-10552092 on Build GRCh38
rs200136327 Chr.19: 10552190-10552190 on Build GRCh38
rs138092907 Chr.19: 10552242-10552242 on Build GRCh38
rs750985644 Chr.19: 10552137-10552137 on Build GRCh38
rs747023526 Chr.19: 10552166-10552166 on Build GRCh38
rs201971548 Chr.19: 10552155-10552155 on Build GRCh38
rs775280853 Chr.19: 10552191-10552191 on Build GRCh38
rs923787724 Chr.19: 10552142-10552142 on Build GRCh38
rs1032804091 Chr.19: 10552237-10552238 on Build GRCh38
rs34986913 Chr.19: 10552111-10552112 on Build GRCh38
rs762841570 Chr.19: 10552125-10552125 on Build GRCh38
rs759152766 Chr.19: 10552138-10552138 on Build GRCh38
rs201728867 Chr.19: 10552237-10552237 on Build GRCh38
rs371466606 Chr.19: 10552227-10552227 on Build GRCh38
rs752118366 Chr.19: 10552222-10552222 on Build GRCh38
rs780044450 Chr.19: 10552067-10552067 on Build GRCh38
rs772157379 Chr.19: 10552061-10552061 on Build GRCh38
rs768822162 Chr.19: 10552094-10552094 on Build GRCh38
rs755043990 Chr.19: 10552224-10552224 on Build GRCh38

View Full Product Details