Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 88166201-88166462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
UniGene
Hs.331268
Gene Chromosome Location
Chr.12: 88142296-88199887 on Build GRCh38
Transcripts
NM_181783.3 XM_011537980.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3466242 c.890C>T p.P297L Chr.12: 88166422-88166422 on Build GRCh38
4575772 c.826A>G p.T276A Chr.12: 88166358-88166358 on Build GRCh38
944255 c.874T>C p.W292R Chr.12: 88166406-88166406 on Build GRCh38
6037596 c.828T>A p.T276T Chr.12: 88166360-88166360 on Build GRCh38
6305199 c.897G>T p.E299D Chr.12: 88166429-88166429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756607630 Chr.12: 88166428-88166428 on Build GRCh38
rs746712283 Chr.12: 88166308-88166308 on Build GRCh38
rs771146399 Chr.12: 88166297-88166297 on Build GRCh38
rs1023864695 Chr.12: 88166397-88166397 on Build GRCh38
rs749951855 Chr.12: 88166349-88166349 on Build GRCh38
rs370059357 Chr.12: 88166360-88166360 on Build GRCh38
rs139775413 Chr.12: 88166357-88166357 on Build GRCh38
rs139374194 Chr.12: 88166402-88166402 on Build GRCh38
rs776671986 Chr.12: 88166315-88166315 on Build GRCh38
rs150884457 Chr.12: 88166365-88166365 on Build GRCh38
rs749577318 Chr.12: 88166293-88166293 on Build GRCh38
rs764900307 Chr.12: 88166339-88166339 on Build GRCh38
rs761822301 Chr.12: 88166324-88166324 on Build GRCh38
rs1052153761 Chr.12: 88166307-88166308 on Build GRCh38
rs760541738 Chr.12: 88166306-88166306 on Build GRCh38
rs781386456 Chr.12: 88166415-88166415 on Build GRCh38
rs935096394 Chr.12: 88166288-88166288 on Build GRCh38
rs763757571 Chr.12: 88166311-88166311 on Build GRCh38
rs911140363 Chr.12: 88166259-88166259 on Build GRCh38
rs755407351 Chr.12: 88166414-88166414 on Build GRCh38
rs1014834431 Chr.12: 88166243-88166243 on Build GRCh38
rs774379456 Chr.12: 88166302-88166302 on Build GRCh38
rs369513030 Chr.12: 88166283-88166283 on Build GRCh38
rs1040889329 Chr.12: 88166277-88166277 on Build GRCh38
rs769908041 Chr.12: 88166282-88166282 on Build GRCh38
rs942535108 Chr.12: 88166269-88166269 on Build GRCh38
rs145643372 Chr.12: 88166427-88166427 on Build GRCh38
rs984073073 Chr.12: 88166253-88166253 on Build GRCh38

View Full Product Details