Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128285878-128286137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SWI5
Entrez Gene ID
375757
Gene Name
SWI5 homologous recombination repair protein
Gene Aliases
C9orf119, SAE3
UniGene
Hs.259594
Gene Chromosome Location
Chr.9: 128275379-128288989 on Build GRCh38
Transcripts
NM_001040011.1 NM_001318089.1 NM_001318092.1 XM_011518652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4731076 c.643G>T p.A215S Chr.9: 128286033-128286033 on Build GRCh38
3654591 c.602A>G p.N201S Chr.9: 128285992-128285992 on Build GRCh38
4425349 c.643+1G>C p.? Chr.9: 128286034-128286034 on Build GRCh38
752636 c.565G>T p.E189* Chr.9: 128285955-128285955 on Build GRCh38
6403367 c.626T>A p.M209K Chr.9: 128286016-128286016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371609493 Chr.9: 128286025-128286025 on Build GRCh38
rs763654836 Chr.9: 128286050-128286050 on Build GRCh38
rs551751889 Chr.9: 128286009-128286009 on Build GRCh38
rs749338568 Chr.9: 128285970-128285970 on Build GRCh38
rs780970947 Chr.9: 128285945-128285945 on Build GRCh38
rs751173468 Chr.9: 128285941-128285941 on Build GRCh38
rs544048008 Chr.9: 128285918-128285918 on Build GRCh38
rs749177303 Chr.9: 128286086-128286086 on Build GRCh38
rs368228872 Chr.9: 128286074-128286074 on Build GRCh38
rs971866265 Chr.9: 128285983-128285983 on Build GRCh38
rs763849782 Chr.9: 128285992-128285992 on Build GRCh38
rs768918042 Chr.9: 128285976-128285976 on Build GRCh38
rs777543502 Chr.9: 128285928-128285928 on Build GRCh38
rs766961317 Chr.9: 128286000-128286000 on Build GRCh38
rs79429554 Chr.9: 128286118-128286118 on Build GRCh38
rs761151635 Chr.9: 128285999-128285999 on Build GRCh38
rs772328672 Chr.9: 128285987-128285987 on Build GRCh38
rs528157671 Chr.9: 128286096-128286096 on Build GRCh38
rs780177680 Chr.9: 128285967-128285967 on Build GRCh38
rs745802668 Chr.9: 128285959-128285959 on Build GRCh38
rs751172309 Chr.9: 128286051-128286051 on Build GRCh38
rs370235235 Chr.9: 128285984-128285984 on Build GRCh38
rs771379352 Chr.9: 128286061-128286061 on Build GRCh38
rs752249975 Chr.9: 128285908-128285908 on Build GRCh38
rs374803657 Chr.9: 128286063-128286063 on Build GRCh38
rs998387858 Chr.9: 128286082-128286082 on Build GRCh38
rs556130465 Chr.9: 128286094-128286095 on Build GRCh38
rs762572083 Chr.9: 128286037-128286037 on Build GRCh38
rs768756659 Chr.9: 128286091-128286091 on Build GRCh38
rs756089316 Chr.9: 128286081-128286081 on Build GRCh38
rs559697777 Chr.9: 128286080-128286080 on Build GRCh38
rs374963645 Chr.9: 128285985-128285985 on Build GRCh38
rs1039959378 Chr.9: 128286072-128286072 on Build GRCh38
rs367776985 Chr.9: 128285994-128285994 on Build GRCh38
rs931574669 Chr.9: 128286093-128286093 on Build GRCh38
rs757031716 Chr.9: 128285944-128285944 on Build GRCh38
rs758163296 Chr.9: 128285927-128285927 on Build GRCh38
rs779948848 Chr.9: 128286085-128286085 on Build GRCh38
rs769753523 Chr.9: 128285966-128285966 on Build GRCh38
rs764853101 Chr.9: 128285903-128285903 on Build GRCh38
rs181886058 Chr.9: 128285958-128285958 on Build GRCh38

View Full Product Details