Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21856783-21857038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBTPS2
Entrez Gene ID
51360
Gene Name
membrane bound transcription factor peptidase, site 2
Gene Aliases
BRESEK, IFAP, KFSD, KFSDX, OLMSX, S2P
UniGene
Hs.443490
Gene Chromosome Location
Chr.X: 21839538-21885423 on Build GRCh38
Transcripts
NM_015884.3 NM_206923.3

Gene Symbol
YY2
Entrez Gene ID
404281
Gene Name
YY2 transcription factor
Gene Aliases
ZNF631
UniGene
Hs.673601
Gene Chromosome Location
Chr.X: 21855987-21858727 on Build GRCh38
Transcripts
NM_015884.3 NM_206923.3

SNP Detail

SNP ID Chromosome Location
rs768424915 Chr.X: 21856866-21856867 on Build GRCh38
rs763777922 Chr.X: 21857007-21857007 on Build GRCh38
rs189927135 Chr.X: 21856816-21856816 on Build GRCh38
rs748893660 Chr.X: 21856841-21856841 on Build GRCh38
rs781656677 Chr.X: 21856808-21856808 on Build GRCh38
rs775288900 Chr.X: 21856976-21856976 on Build GRCh38
rs774055284 Chr.X: 21856867-21856872 on Build GRCh38
rs748868186 Chr.X: 21856941-21856941 on Build GRCh38
rs781700906 Chr.X: 21856920-21856920 on Build GRCh38
rs757318606 Chr.X: 21856988-21856988 on Build GRCh38
rs773806383 Chr.X: 21856952-21856952 on Build GRCh38
rs138260436 Chr.X: 21856942-21856942 on Build GRCh38
rs761456434 Chr.X: 21856895-21856906 on Build GRCh38
rs371573327 Chr.X: 21856864-21856864 on Build GRCh38
rs766537345 Chr.X: 21856898-21856898 on Build GRCh38
rs772601524 Chr.X: 21856828-21856828 on Build GRCh38
rs755494527 Chr.X: 21856919-21856919 on Build GRCh38
rs777538519 Chr.X: 21856924-21856924 on Build GRCh38
rs747374702 Chr.X: 21856865-21856865 on Build GRCh38
rs780275038 Chr.X: 21857018-21857018 on Build GRCh38
rs763229180 Chr.X: 21857010-21857010 on Build GRCh38
rs767015419 Chr.X: 21856855-21856855 on Build GRCh38
rs891357357 Chr.X: 21856832-21856832 on Build GRCh38
rs752412245 Chr.X: 21856858-21856858 on Build GRCh38
rs767326908 Chr.X: 21856861-21856861 on Build GRCh38
rs752145497 Chr.X: 21856901-21856901 on Build GRCh38
rs773525579 Chr.X: 21857008-21857008 on Build GRCh38
rs371278958 Chr.X: 21856964-21856964 on Build GRCh38
rs571331916 Chr.X: 21856922-21856922 on Build GRCh38
rs747779901 Chr.X: 21856815-21856815 on Build GRCh38
rs750600223 Chr.X: 21856878-21856878 on Build GRCh38
rs755928626 Chr.X: 21856862-21856862 on Build GRCh38
rs770412899 Chr.X: 21856847-21856847 on Build GRCh38
rs777747632 Chr.X: 21856886-21856886 on Build GRCh38
rs867019385 Chr.X: 21856875-21856875 on Build GRCh38
rs374425110 Chr.X: 21856923-21856923 on Build GRCh38
rs367599474 Chr.X: 21856955-21856955 on Build GRCh38
rs891364185 Chr.X: 21856981-21856981 on Build GRCh38

View Full Product Details