Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52461010-52461267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2
Entrez Gene ID
26586
Gene Name
cytoskeleton associated protein 2
Gene Aliases
LB1, TMAP, se20-10
UniGene
Hs.444028
Gene Chromosome Location
Chr.13: 52455360-52476631 on Build GRCh38
Transcripts
NM_001098525.2 NM_001286686.1 NM_001286687.1 NM_018204.4 XM_005266344.4 XM_011535043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575982 c.412A>G p.S138G Chr.13: 52461238-52461238 on Build GRCh38
4617473 c.381G>A p.P127P Chr.13: 52461207-52461207 on Build GRCh38
5376831 c.302C>T p.P101L Chr.13: 52461128-52461128 on Build GRCh38
5121468 c.437_1305+420>25 p.? Chr.13: 52461263-52462987 on Build GRCh38
5842647 c.406C>T p.P136S Chr.13: 52461232-52461232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750701904 Chr.13: 52461101-52461101 on Build GRCh38
rs373545539 Chr.13: 52461064-52461064 on Build GRCh38
rs36022164 Chr.13: 52461155-52461155 on Build GRCh38
rs376367571 Chr.13: 52461093-52461093 on Build GRCh38
rs917946986 Chr.13: 52461227-52461227 on Build GRCh38
rs764723599 Chr.13: 52461066-52461066 on Build GRCh38
rs539183420 Chr.13: 52461074-52461074 on Build GRCh38
rs746670021 Chr.13: 52461191-52461191 on Build GRCh38
rs777774781 Chr.13: 52461161-52461161 on Build GRCh38
rs772813318 Chr.13: 52461180-52461180 on Build GRCh38
rs767832969 Chr.13: 52461084-52461084 on Build GRCh38
rs776001339 Chr.13: 52461058-52461058 on Build GRCh38
rs566498056 Chr.13: 52461054-52461054 on Build GRCh38
rs763445264 Chr.13: 52461207-52461207 on Build GRCh38
rs979583979 Chr.13: 52461122-52461122 on Build GRCh38
rs546281774 Chr.13: 52461092-52461092 on Build GRCh38
rs771592179 Chr.13: 52461163-52461163 on Build GRCh38
rs764480140 Chr.13: 52461225-52461225 on Build GRCh38
rs149189169 Chr.13: 52461120-52461120 on Build GRCh38
rs774976799 Chr.13: 52461230-52461230 on Build GRCh38
rs775705785 Chr.13: 52461206-52461206 on Build GRCh38
rs778948021 Chr.13: 52461142-52461142 on Build GRCh38
rs779723818 Chr.13: 52461121-52461121 on Build GRCh38
rs749162412 Chr.13: 52461137-52461137 on Build GRCh38
rs772899454 Chr.13: 52461040-52461040 on Build GRCh38
rs138583090 Chr.13: 52461205-52461205 on Build GRCh38
rs771681941 Chr.13: 52461039-52461039 on Build GRCh38
rs369526042 Chr.13: 52461055-52461055 on Build GRCh38
rs770006252 Chr.13: 52461131-52461132 on Build GRCh38
rs758498329 Chr.13: 52461141-52461141 on Build GRCh38
rs778359504 Chr.13: 52461173-52461174 on Build GRCh38
rs925451353 Chr.13: 52461129-52461129 on Build GRCh38
rs757457224 Chr.13: 52461082-52461082 on Build GRCh38

View Full Product Details