Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2172314-2172555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF7
Entrez Gene ID
84231
Gene Name
TNF receptor associated factor 7
Gene Aliases
RFWD1, RNF119
UniGene
Hs.713632
Gene Chromosome Location
Chr.16: 2155752-2178129 on Build GRCh38
Transcripts
NM_032271.2 XM_005255627.4 XM_011522700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172738 c.636C>T p.F212F Chr.16: 2172351-2172351 on Build GRCh38
6189813 c.616G>C p.D206H Chr.16: 2172331-2172331 on Build GRCh38
6973563 c.623G>A p.R208Q Chr.16: 2172338-2172338 on Build GRCh38
434892 c.746C>A p.A249D Chr.16: 2172551-2172551 on Build GRCh38
5384504 c.618C>T p.D206D Chr.16: 2172333-2172333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768713660 Chr.16: 2172466-2172466 on Build GRCh38
rs766165474 Chr.16: 2172499-2172499 on Build GRCh38
rs771191963 Chr.16: 2172338-2172338 on Build GRCh38
rs764852847 Chr.16: 2172522-2172522 on Build GRCh38
rs764764982 Chr.16: 2172431-2172431 on Build GRCh38
rs775571912 Chr.16: 2172364-2172364 on Build GRCh38
rs914029813 Chr.16: 2172371-2172371 on Build GRCh38
rs888344660 Chr.16: 2172460-2172460 on Build GRCh38
rs761727088 Chr.16: 2172422-2172422 on Build GRCh38
rs749436466 Chr.16: 2172459-2172459 on Build GRCh38
rs780219733 Chr.16: 2172453-2172453 on Build GRCh38
rs751996644 Chr.16: 2172439-2172439 on Build GRCh38
rs748044013 Chr.16: 2172468-2172468 on Build GRCh38
rs772902058 Chr.16: 2172474-2172474 on Build GRCh38
rs757728472 Chr.16: 2172527-2172527 on Build GRCh38
rs182666045 Chr.16: 2172452-2172452 on Build GRCh38
rs370331170 Chr.16: 2172424-2172424 on Build GRCh38
rs760492852 Chr.16: 2172492-2172492 on Build GRCh38
rs781530832 Chr.16: 2172340-2172340 on Build GRCh38
rs556428097 Chr.16: 2172427-2172427 on Build GRCh38
rs375174789 Chr.16: 2172519-2172519 on Build GRCh38
rs776414915 Chr.16: 2172500-2172500 on Build GRCh38
rs768623972 Chr.16: 2172367-2172367 on Build GRCh38
rs769966564 Chr.16: 2172357-2172357 on Build GRCh38
rs765716964 Chr.16: 2172382-2172382 on Build GRCh38
rs564325600 Chr.16: 2172528-2172528 on Build GRCh38
rs746197336 Chr.16: 2172446-2172446 on Build GRCh38
rs750793648 Chr.16: 2172532-2172532 on Build GRCh38
rs749350622 Chr.16: 2172366-2172366 on Build GRCh38
rs141784925 Chr.16: 2172342-2172342 on Build GRCh38
rs750035555 Chr.16: 2172522-2172522 on Build GRCh38
rs760274832 Chr.16: 2172426-2172426 on Build GRCh38
rs1055301267 Chr.16: 2172531-2172531 on Build GRCh38
rs910346978 Chr.16: 2172448-2172448 on Build GRCh38
rs1015601872 Chr.16: 2172361-2172361 on Build GRCh38
rs781413009 Chr.16: 2172445-2172445 on Build GRCh38
rs752183367 Chr.16: 2172525-2172525 on Build GRCh38
rs757722846 Chr.16: 2172443-2172443 on Build GRCh38
rs544428296 Chr.16: 2172471-2172471 on Build GRCh38
rs774267754 Chr.16: 2172394-2172394 on Build GRCh38

View Full Product Details