Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43922072-43922345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF45
Entrez Gene ID
7596
Gene Name
zinc finger protein 45
Gene Aliases
KOX5, ZNF13
UniGene
Hs.381285
Gene Chromosome Location
Chr.19: 43912624-43935278 on Build GRCh38
Transcripts
NM_003425.3 XM_011527267.1 XM_011527269.1 XM_011527271.1 XM_011527273.1 XM_017027217.1 XM_017027218.1 XM_017027219.1 XM_017027220.1 XM_017027221.1 XM_017027222.1 XM_017027223.1 XM_017027224.1 XM_017027225.1 XM_017027226.1 XM_017027227.1 XM_017027228.1 XM_017027229.1 XM_017027230.1

SNP Detail

SNP ID Chromosome Location
rs113258371 Chr.19: 43922215-43922215 on Build GRCh38
rs551378009 Chr.19: 43922180-43922180 on Build GRCh38
rs765465758 Chr.19: 43922137-43922137 on Build GRCh38
rs759678466 Chr.19: 43922166-43922166 on Build GRCh38
rs761566660 Chr.19: 43922134-43922134 on Build GRCh38
rs188136163 Chr.19: 43922190-43922190 on Build GRCh38
rs767586540 Chr.19: 43922174-43922174 on Build GRCh38
rs201533198 Chr.19: 43922311-43922312 on Build GRCh38
rs1005707020 Chr.19: 43922273-43922273 on Build GRCh38
rs1006910132 Chr.19: 43922311-43922311 on Build GRCh38
rs377288497 Chr.19: 43922140-43922140 on Build GRCh38
rs973921425 Chr.19: 43922111-43922111 on Build GRCh38
rs75063630 Chr.19: 43922315-43922315 on Build GRCh38
rs955030196 Chr.19: 43922307-43922315 on Build GRCh38
rs772793780 Chr.19: 43922312-43922315 on Build GRCh38
rs200912684 Chr.19: 43922192-43922192 on Build GRCh38
rs57712519 Chr.19: 43922315-43922316 on Build GRCh38
rs1017042108 Chr.19: 43922274-43922275 on Build GRCh38
rs375536228 Chr.19: 43922274-43922274 on Build GRCh38
rs373181320 Chr.19: 43922125-43922125 on Build GRCh38
rs891954955 Chr.19: 43922206-43922206 on Build GRCh38
rs413093 Chr.19: 43922113-43922113 on Build GRCh38
rs1054557559 Chr.19: 43922203-43922203 on Build GRCh38
rs751571025 Chr.19: 43922157-43922157 on Build GRCh38
rs891148443 Chr.19: 43922300-43922300 on Build GRCh38
rs756350135 Chr.19: 43922181-43922181 on Build GRCh38
rs866988248 Chr.19: 43922249-43922249 on Build GRCh38
rs1018266048 Chr.19: 43922312-43922312 on Build GRCh38
rs562297912 Chr.19: 43922105-43922105 on Build GRCh38
rs780094225 Chr.19: 43922209-43922209 on Build GRCh38
rs768413256 Chr.19: 43922126-43922126 on Build GRCh38
rs1029895458 Chr.19: 43922288-43922288 on Build GRCh38
rs1044612062 Chr.19: 43922232-43922232 on Build GRCh38
rs66834021 Chr.19: 43922316-43922317 on Build GRCh38
rs1010944502 Chr.19: 43922276-43922276 on Build GRCh38
rs7246971 Chr.19: 43922316-43922316 on Build GRCh38
rs184394406 Chr.19: 43922108-43922108 on Build GRCh38
rs906105904 Chr.19: 43922261-43922261 on Build GRCh38
rs985971408 Chr.19: 43922100-43922100 on Build GRCh38
rs200142032 Chr.19: 43922148-43922148 on Build GRCh38
rs371101091 Chr.19: 43922147-43922147 on Build GRCh38
rs200820901 Chr.19: 43922311-43922311 on Build GRCh38
rs962759573 Chr.19: 43922109-43922109 on Build GRCh38
rs1029295090 Chr.19: 43922294-43922296 on Build GRCh38
rs757264288 Chr.19: 43922223-43922240 on Build GRCh38
rs547404256 Chr.19: 43922256-43922256 on Build GRCh38

View Full Product Details