Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163471714-163471959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TKTL2
Entrez Gene ID
84076
Gene Name
transketolase like 2
Gene Aliases
-
UniGene
Hs.303923
Gene Chromosome Location
Chr.4: 163471095-163473895 on Build GRCh38
Transcripts
NM_032136.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1052864 c.1856C>T p.A619V Chr.4: 163471879-163471879 on Build GRCh38
323833 c.1819G>C p.D607H Chr.4: 163471916-163471916 on Build GRCh38
6529376 c.1864C>G p.L622V Chr.4: 163471871-163471871 on Build GRCh38
6802130 c.1824G>A p.M608I Chr.4: 163471911-163471911 on Build GRCh38
1052863 c.1858G>A p.V620I Chr.4: 163471877-163471877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749187159 Chr.4: 163471817-163471817 on Build GRCh38
rs756094486 Chr.4: 163471806-163471806 on Build GRCh38
rs139967072 Chr.4: 163471861-163471861 on Build GRCh38
rs201590203 Chr.4: 163471929-163471929 on Build GRCh38
rs768231270 Chr.4: 163471867-163471867 on Build GRCh38
rs773623685 Chr.4: 163471878-163471878 on Build GRCh38
rs768081131 Chr.4: 163471905-163471905 on Build GRCh38
rs767522920 Chr.4: 163471887-163471887 on Build GRCh38
rs746502311 Chr.4: 163471864-163471864 on Build GRCh38
rs778636827 Chr.4: 163471917-163471917 on Build GRCh38
rs766403360 Chr.4: 163471822-163471822 on Build GRCh38
rs754770325 Chr.4: 163471880-163471880 on Build GRCh38
rs762343590 Chr.4: 163471874-163471874 on Build GRCh38
rs757053805 Chr.4: 163471838-163471838 on Build GRCh38
rs151077178 Chr.4: 163471883-163471883 on Build GRCh38
rs145457799 Chr.4: 163471877-163471877 on Build GRCh38
rs777307971 Chr.4: 163471871-163471871 on Build GRCh38
rs541496760 Chr.4: 163471939-163471939 on Build GRCh38
rs771746721 Chr.4: 163471849-163471849 on Build GRCh38
rs746471827 Chr.4: 163471882-163471882 on Build GRCh38
rs745561380 Chr.4: 163471844-163471844 on Build GRCh38
rs779776530 Chr.4: 163471940-163471940 on Build GRCh38
rs970258503 Chr.4: 163471812-163471812 on Build GRCh38
rs751499122 Chr.4: 163471834-163471834 on Build GRCh38
rs548320199 Chr.4: 163471782-163471782 on Build GRCh38
rs759645946 Chr.4: 163471885-163471885 on Build GRCh38
rs757169252 Chr.4: 163471916-163471916 on Build GRCh38
rs751685516 Chr.4: 163471884-163471884 on Build GRCh38
rs376040167 Chr.4: 163471828-163471828 on Build GRCh38
rs778715163 Chr.4: 163471843-163471843 on Build GRCh38
rs753790590 Chr.4: 163471906-163471906 on Build GRCh38

View Full Product Details