Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28023264-28023517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUGGC
Entrez Gene ID
389643
Gene Name
nuclear GTPase, germinal center associated
Gene Aliases
C8orf80, HMFN0672, SLIP-GC
UniGene
Hs.370129
Gene Chromosome Location
Chr.8: 28021964-28084068 on Build GRCh38
Transcripts
NM_001010906.1 XM_011544523.2 XM_011544524.2 XM_011544525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098806 c.2335G>T p.E779* Chr.8: 28023373-28023373 on Build GRCh38
5119992 c.2307G>T p.E769D Chr.8: 28023401-28023401 on Build GRCh38
3647878 c.2377G>C p.G793R Chr.8: 28023331-28023331 on Build GRCh38
6261407 c.2361C>A p.S787R Chr.8: 28023347-28023347 on Build GRCh38
3647882 c.2355C>T p.S785S Chr.8: 28023353-28023353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535677411 Chr.8: 28023373-28023373 on Build GRCh38
rs775199896 Chr.8: 28023314-28023314 on Build GRCh38
rs945869518 Chr.8: 28023448-28023448 on Build GRCh38
rs750907742 Chr.8: 28023333-28023333 on Build GRCh38
rs762210589 Chr.8: 28023396-28023396 on Build GRCh38
rs760299604 Chr.8: 28023315-28023315 on Build GRCh38
rs771946255 Chr.8: 28023478-28023478 on Build GRCh38
rs745798993 Chr.8: 28023471-28023471 on Build GRCh38
rs149363017 Chr.8: 28023351-28023351 on Build GRCh38
rs759033231 Chr.8: 28023299-28023299 on Build GRCh38
rs549505578 Chr.8: 28023394-28023394 on Build GRCh38
rs369859443 Chr.8: 28023354-28023354 on Build GRCh38
rs771844076 Chr.8: 28023332-28023332 on Build GRCh38
rs749037056 Chr.8: 28023339-28023339 on Build GRCh38
rs770729694 Chr.8: 28023341-28023341 on Build GRCh38
rs535303167 Chr.8: 28023433-28023433 on Build GRCh38
rs921597 Chr.8: 28023309-28023309 on Build GRCh38
rs781426854 Chr.8: 28023338-28023338 on Build GRCh38
rs949341543 Chr.8: 28023432-28023432 on Build GRCh38
rs200969254 Chr.8: 28023480-28023480 on Build GRCh38
rs771905699 Chr.8: 28023331-28023332 on Build GRCh38
rs778626833 Chr.8: 28023466-28023466 on Build GRCh38
rs201330243 Chr.8: 28023337-28023337 on Build GRCh38
rs774298309 Chr.8: 28023297-28023297 on Build GRCh38
rs745855698 Chr.8: 28023348-28023348 on Build GRCh38
rs376549983 Chr.8: 28023497-28023497 on Build GRCh38
rs770817728 Chr.8: 28023296-28023296 on Build GRCh38
rs763797956 Chr.8: 28023393-28023393 on Build GRCh38
rs941779047 Chr.8: 28023368-28023368 on Build GRCh38
rs189541232 Chr.8: 28023334-28023334 on Build GRCh38
rs768294923 Chr.8: 28023487-28023487 on Build GRCh38
rs760341571 Chr.8: 28023382-28023382 on Build GRCh38
rs376205193 Chr.8: 28023417-28023417 on Build GRCh38
rs757038145 Chr.8: 28023457-28023457 on Build GRCh38
rs907639735 Chr.8: 28023361-28023361 on Build GRCh38
rs749195429 Chr.8: 28023292-28023292 on Build GRCh38
rs765700940 Chr.8: 28023404-28023404 on Build GRCh38
rs112798378 Chr.8: 28023406-28023406 on Build GRCh38
rs561844649 Chr.8: 28023329-28023329 on Build GRCh38
rs181080274 Chr.8: 28023490-28023490 on Build GRCh38
rs752925594 Chr.8: 28023453-28023453 on Build GRCh38
rs751556152 Chr.8: 28023427-28023427 on Build GRCh38
rs370883557 Chr.8: 28023489-28023489 on Build GRCh38
rs781337018 Chr.8: 28023446-28023446 on Build GRCh38
rs982876365 Chr.8: 28023360-28023360 on Build GRCh38
rs773041506 Chr.8: 28023285-28023285 on Build GRCh38
rs376263360 Chr.8: 28023331-28023331 on Build GRCh38
rs372657420 Chr.8: 28023343-28023343 on Build GRCh38
rs377609860 Chr.8: 28023405-28023405 on Build GRCh38
rs746470781 Chr.8: 28023481-28023481 on Build GRCh38
rs1052130786 Chr.8: 28023407-28023407 on Build GRCh38

View Full Product Details