Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15187910-15188181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39B
Entrez Gene ID
158219
Gene Name
tetratricopeptide repeat domain 39B
Gene Aliases
C9orf52
UniGene
Hs.563630
Gene Chromosome Location
Chr.9: 15167204-15307360 on Build GRCh38
Transcripts
NM_001168339.1 NM_001168340.1 NM_001168341.1 NM_001168342.1 NM_152574.2 XM_011517732.2 XM_017014310.1 XM_017014311.1 XM_017014312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4999748 c.1354T>C p.Y452H Chr.9: 15188012-15188012 on Build GRCh38
455697 c.1258A>G p.I420V Chr.9: 15188108-15188108 on Build GRCh38
4524543 c.1278G>A p.W426* Chr.9: 15188088-15188088 on Build GRCh38
1598200 c.1296C>A p.L432L Chr.9: 15188070-15188070 on Build GRCh38
7309274 c.1264T>A p.W422R Chr.9: 15188063-15188063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151093046 Chr.9: 15188091-15188091 on Build GRCh38
rs777340350 Chr.9: 15187938-15187938 on Build GRCh38
rs372841195 Chr.9: 15187974-15187974 on Build GRCh38
rs759548750 Chr.9: 15188079-15188079 on Build GRCh38
rs185198420 Chr.9: 15188136-15188136 on Build GRCh38
rs776436527 Chr.9: 15188080-15188080 on Build GRCh38
rs776488537 Chr.9: 15187986-15187986 on Build GRCh38
rs146887179 Chr.9: 15188115-15188115 on Build GRCh38
rs576572202 Chr.9: 15188099-15188099 on Build GRCh38
rs776473593 Chr.9: 15188033-15188033 on Build GRCh38
rs150371121 Chr.9: 15188020-15188020 on Build GRCh38
rs1048200174 Chr.9: 15188068-15188068 on Build GRCh38
rs190605572 Chr.9: 15187963-15187963 on Build GRCh38
rs377546314 Chr.9: 15188135-15188135 on Build GRCh38
rs773487358 Chr.9: 15187978-15187978 on Build GRCh38
rs762412435 Chr.9: 15188007-15188007 on Build GRCh38
rs757448800 Chr.9: 15188149-15188149 on Build GRCh38
rs192997470 Chr.9: 15188109-15188109 on Build GRCh38
rs756479249 Chr.9: 15188024-15188024 on Build GRCh38
rs771815650 Chr.9: 15187982-15187982 on Build GRCh38
rs371262247 Chr.9: 15188097-15188097 on Build GRCh38
rs762513609 Chr.9: 15188093-15188093 on Build GRCh38
rs765758498 Chr.9: 15188129-15188129 on Build GRCh38
rs754347153 Chr.9: 15188033-15188033 on Build GRCh38
rs779324290 Chr.9: 15188043-15188043 on Build GRCh38
rs758462716 Chr.9: 15188050-15188050 on Build GRCh38
rs746126448 Chr.9: 15188096-15188096 on Build GRCh38
rs777864932 Chr.9: 15188147-15188147 on Build GRCh38
rs756616066 Chr.9: 15187947-15187947 on Build GRCh38
rs763837031 Chr.9: 15188014-15188014 on Build GRCh38
rs377562656 Chr.9: 15188151-15188151 on Build GRCh38
rs757142605 Chr.9: 15188037-15188037 on Build GRCh38
rs895435267 Chr.9: 15188104-15188104 on Build GRCh38
rs759553167 Chr.9: 15187991-15187991 on Build GRCh38
rs142900421 Chr.9: 15188066-15188066 on Build GRCh38
rs1020134169 Chr.9: 15188132-15188132 on Build GRCh38
rs982980815 Chr.9: 15188008-15188008 on Build GRCh38
rs74561491 Chr.9: 15187994-15187994 on Build GRCh38
rs141394265 Chr.9: 15188095-15188095 on Build GRCh38
rs919724595 Chr.9: 15187993-15187993 on Build GRCh38
rs745700070 Chr.9: 15188082-15188082 on Build GRCh38
rs760447227 Chr.9: 15187980-15187980 on Build GRCh38
rs750787676 Chr.9: 15187945-15187945 on Build GRCh38
rs376401966 Chr.9: 15188028-15188028 on Build GRCh38
rs973689966 Chr.9: 15187975-15187975 on Build GRCh38
rs748221856 Chr.9: 15187970-15187970 on Build GRCh38
rs752230423 Chr.9: 15188005-15188005 on Build GRCh38
rs543379562 Chr.9: 15188058-15188058 on Build GRCh38
rs749816683 Chr.9: 15187957-15187957 on Build GRCh38
rs780647276 Chr.9: 15187954-15187954 on Build GRCh38
rs755484760 Chr.9: 15188034-15188034 on Build GRCh38
rs768577731 Chr.9: 15187961-15187961 on Build GRCh38
rs1037492403 Chr.9: 15188017-15188017 on Build GRCh38
rs777916444 Chr.9: 15188055-15188055 on Build GRCh38
rs944057059 Chr.9: 15187998-15187998 on Build GRCh38
rs144921032 Chr.9: 15188048-15188048 on Build GRCh38
rs1407977 Chr.9: 15188108-15188108 on Build GRCh38

View Full Product Details