Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39459444-39459705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT5H
Entrez Gene ID
6829
Gene Name
SPT5 homolog, DSIF elongation factor subunit
Gene Aliases
SPT5, SPT5H, Tat-CT1
UniGene
Hs.631604
Gene Chromosome Location
Chr.19: 39445546-39476670 on Build GRCh38
Transcripts
NM_001111020.2 NM_001130824.1 NM_001130825.1 NM_001319990.1 NM_001319991.1 NM_003169.3 XM_017027174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
711479 c.546C>G p.V182V Chr.19: 39459580-39459580 on Build GRCh38
1393628 c.527A>T p.D176V Chr.19: 39459561-39459561 on Build GRCh38
5543490 c.531C>T p.P177P Chr.19: 39459565-39459565 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754008995 Chr.19: 39459606-39459606 on Build GRCh38
rs1130184 Chr.19: 39459576-39459576 on Build GRCh38
rs144810638 Chr.19: 39459613-39459613 on Build GRCh38
rs909490685 Chr.19: 39459674-39459674 on Build GRCh38
rs745469514 Chr.19: 39459540-39459540 on Build GRCh38
rs909776304 Chr.19: 39459484-39459484 on Build GRCh38
rs188234827 Chr.19: 39459661-39459661 on Build GRCh38
rs746772200 Chr.19: 39459510-39459510 on Build GRCh38
rs180860273 Chr.19: 39459476-39459476 on Build GRCh38
rs746862455 Chr.19: 39459629-39459629 on Build GRCh38
rs1031430292 Chr.19: 39459495-39459495 on Build GRCh38
rs369098114 Chr.19: 39459598-39459598 on Build GRCh38
rs761119385 Chr.19: 39459595-39459595 on Build GRCh38
rs773755743 Chr.19: 39459589-39459589 on Build GRCh38
rs758371189 Chr.19: 39459625-39459625 on Build GRCh38
rs1012892598 Chr.19: 39459600-39459600 on Build GRCh38
rs969581202 Chr.19: 39459648-39459648 on Build GRCh38
rs553517277 Chr.19: 39459680-39459680 on Build GRCh38
rs985352406 Chr.19: 39459482-39459482 on Build GRCh38
rs999559952 Chr.19: 39459480-39459480 on Build GRCh38
rs1057452727 Chr.19: 39459550-39459550 on Build GRCh38
rs370030406 Chr.19: 39459519-39459519 on Build GRCh38
rs534847534 Chr.19: 39459672-39459672 on Build GRCh38
rs929400093 Chr.19: 39459528-39459528 on Build GRCh38
rs775028339 Chr.19: 39459565-39459565 on Build GRCh38
rs757055436 Chr.19: 39459638-39459638 on Build GRCh38
rs1036809141 Chr.19: 39459499-39459499 on Build GRCh38
rs866337771 Chr.19: 39459498-39459498 on Build GRCh38
rs776132750 Chr.19: 39459520-39459520 on Build GRCh38
rs552291679 Chr.19: 39459607-39459607 on Build GRCh38
rs904756951 Chr.19: 39459679-39459679 on Build GRCh38
rs374742976 Chr.19: 39459567-39459567 on Build GRCh38
rs780975236 Chr.19: 39459641-39459641 on Build GRCh38
rs142546168 Chr.19: 39459569-39459569 on Build GRCh38
rs186062583 Chr.19: 39459556-39459556 on Build GRCh38
rs985363310 Chr.19: 39459670-39459670 on Build GRCh38

View Full Product Details