Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52435280-52435539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC11
Entrez Gene ID
23318
Gene Name
zinc finger CCHC-type containing 11
Gene Aliases
PAPD3, TUT4
UniGene
Hs.655407
Gene Chromosome Location
Chr.1: 52423275-52553463 on Build GRCh38
Transcripts
NM_001009881.2 NM_015269.2 XM_005270676.2 XM_005270678.2 XM_005270679.2 XM_005270680.2 XM_005270681.2 XM_005270682.2 XM_005270683.2 XM_006710498.2 XM_006710499.3 XM_006710500.1 XM_011541097.1 XM_011541098.1 XM_011541099.1 XM_017000803.1 XM_017000804.1 XM_017000805.1 XM_017000806.1 XM_017000807.1 XM_017000808.1 XM_017000809.1 XM_017000810.1 XM_017000811.1 XM_017000812.1 XM_017000813.1 XM_017000814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6126960 c.4177_4178CG>TT p.R1393F Chr.1: 52435450-52435451 on Build GRCh38
6063713 c.4169A>T p.Q1390L Chr.1: 52435459-52435459 on Build GRCh38
1474069 c.4195C>G p.Q1399E Chr.1: 52435433-52435433 on Build GRCh38
6063714 c.4195C>T p.Q1399* Chr.1: 52435433-52435433 on Build GRCh38
910671 c.4181A>G p.N1394S Chr.1: 52435447-52435447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773285366 Chr.1: 52435472-52435472 on Build GRCh38
rs755627960 Chr.1: 52435446-52435446 on Build GRCh38
rs765358937 Chr.1: 52435509-52435509 on Build GRCh38
rs757429088 Chr.1: 52435450-52435450 on Build GRCh38
rs200195311 Chr.1: 52435447-52435447 on Build GRCh38
rs180941729 Chr.1: 52435366-52435366 on Build GRCh38
rs747916893 Chr.1: 52435461-52435461 on Build GRCh38
rs752024382 Chr.1: 52435444-52435444 on Build GRCh38
rs765695429 Chr.1: 52435411-52435411 on Build GRCh38
rs370900957 Chr.1: 52435367-52435367 on Build GRCh38
rs764611195 Chr.1: 52435434-52435434 on Build GRCh38
rs776842125 Chr.1: 52435498-52435498 on Build GRCh38
rs759660052 Chr.1: 52435404-52435404 on Build GRCh38
rs748875411 Chr.1: 52435341-52435341 on Build GRCh38
rs779715790 Chr.1: 52435334-52435334 on Build GRCh38
rs759846808 Chr.1: 52435508-52435508 on Build GRCh38
rs771858794 Chr.1: 52435470-52435470 on Build GRCh38
rs752977814 Chr.1: 52435423-52435423 on Build GRCh38
rs773233969 Chr.1: 52435383-52435383 on Build GRCh38
rs938690698 Chr.1: 52435492-52435492 on Build GRCh38
rs748157552 Chr.1: 52435378-52435378 on Build GRCh38
rs771189147 Chr.1: 52435489-52435489 on Build GRCh38
rs777734607 Chr.1: 52435391-52435391 on Build GRCh38
rs547908613 Chr.1: 52435479-52435479 on Build GRCh38
rs758996610 Chr.1: 52435428-52435428 on Build GRCh38
rs956202106 Chr.1: 52435353-52435353 on Build GRCh38
rs938464134 Chr.1: 52435398-52435398 on Build GRCh38
rs771881019 Chr.1: 52435381-52435381 on Build GRCh38
rs777003625 Chr.1: 52435396-52435396 on Build GRCh38
rs143272455 Chr.1: 52435451-52435451 on Build GRCh38

View Full Product Details