Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73983906-73984165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOML1
Entrez Gene ID
9399
Gene Name
stomatin like 1
Gene Aliases
SLP-1, STORP, hUNC-24
UniGene
Hs.194816
Gene Chromosome Location
Chr.15: 73983218-73994622 on Build GRCh38
Transcripts
NM_001256672.1 NM_001256673.1 NM_001256674.1 NM_001256675.1 NM_001256676.1 NM_001256677.1 NM_001324226.1 NM_001324227.1 NM_001324228.1 NM_001324229.1 NM_001324230.1 NM_004809.4 XM_005254792.4 XM_006720773.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982982 c.1182C>T p.L394L Chr.15: 73983952-73983952 on Build GRCh38
6792180 c.1130G>A p.R377Q Chr.15: 73984004-73984004 on Build GRCh38
5849739 c.1103G>A p.R368Q Chr.15: 73984031-73984031 on Build GRCh38
4958020 c.1038C>G p.G346G Chr.15: 73984096-73984096 on Build GRCh38
3816708 c.1059G>A p.E353E Chr.15: 73984075-73984075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749284141 Chr.15: 73984020-73984020 on Build GRCh38
rs756739561 Chr.15: 73984069-73984069 on Build GRCh38
rs772283912 Chr.15: 73984027-73984027 on Build GRCh38
rs1037284716 Chr.15: 73984054-73984054 on Build GRCh38
rs1061082 Chr.15: 73984028-73984028 on Build GRCh38
rs760990889 Chr.15: 73984034-73984034 on Build GRCh38
rs751015227 Chr.15: 73984068-73984068 on Build GRCh38
rs530583189 Chr.15: 73984073-73984073 on Build GRCh38
rs769286917 Chr.15: 73984020-73984020 on Build GRCh38
rs750137997 Chr.15: 73984093-73984093 on Build GRCh38
rs924441562 Chr.15: 73984003-73984003 on Build GRCh38
rs751159750 Chr.15: 73983987-73983987 on Build GRCh38
rs773393667 Chr.15: 73984136-73984136 on Build GRCh38
rs867114873 Chr.15: 73983933-73983933 on Build GRCh38
rs921649345 Chr.15: 73983972-73983972 on Build GRCh38
rs957727690 Chr.15: 73983941-73983941 on Build GRCh38
rs376669719 Chr.15: 73983988-73983988 on Build GRCh38
rs368495173 Chr.15: 73984056-73984056 on Build GRCh38
rs558856616 Chr.15: 73984004-73984004 on Build GRCh38
rs745408599 Chr.15: 73984070-73984070 on Build GRCh38
rs145488707 Chr.15: 73983989-73983989 on Build GRCh38
rs915568499 Chr.15: 73984015-73984015 on Build GRCh38
rs559659064 Chr.15: 73984128-73984128 on Build GRCh38
rs780107270 Chr.15: 73984014-73984014 on Build GRCh38
rs146505992 Chr.15: 73984032-73984032 on Build GRCh38
rs755948842 Chr.15: 73984107-73984107 on Build GRCh38
rs576015466 Chr.15: 73983938-73983938 on Build GRCh38
rs754872992 Chr.15: 73984121-73984121 on Build GRCh38
rs757818032 Chr.15: 73983963-73983963 on Build GRCh38
rs537166637 Chr.15: 73983946-73983946 on Build GRCh38
rs766772797 Chr.15: 73984045-73984045 on Build GRCh38
rs773075698 Chr.15: 73984077-73984078 on Build GRCh38
rs777393936 Chr.15: 73983978-73983978 on Build GRCh38
rs989218573 Chr.15: 73983964-73983964 on Build GRCh38
rs148196033 Chr.15: 73984085-73984085 on Build GRCh38
rs761044311 Chr.15: 73983931-73983931 on Build GRCh38
rs141134528 Chr.15: 73984035-73984035 on Build GRCh38
rs371570677 Chr.15: 73984005-73984005 on Build GRCh38
rs761799346 Chr.15: 73984025-73984025 on Build GRCh38
rs768602137 Chr.15: 73984021-73984021 on Build GRCh38
rs774553753 Chr.15: 73984023-73984023 on Build GRCh38
rs749229006 Chr.15: 73984111-73984111 on Build GRCh38
rs772078974 Chr.15: 73984133-73984133 on Build GRCh38
rs143002933 Chr.15: 73984055-73984055 on Build GRCh38
rs754322978 Chr.15: 73984048-73984048 on Build GRCh38
rs141171144 Chr.15: 73984110-73984110 on Build GRCh38

View Full Product Details