Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56401656-56401921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMFR
Entrez Gene ID
267
Gene Name
autocrine motility factor receptor
Gene Aliases
GP78, RNF45
UniGene
Hs.295137
Gene Chromosome Location
Chr.16: 56361452-56425538 on Build GRCh38
Transcripts
NM_001144.5 NM_001323511.1 NM_001323512.1 XM_005255890.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799056 c.1083C>G p.H361Q Chr.16: 56401735-56401735 on Build GRCh38
4881355 c.1080C>A p.F360L Chr.16: 56401738-56401738 on Build GRCh38
1324395 c.1060C>G p.L354V Chr.16: 56401758-56401758 on Build GRCh38
1182672 c.1055G>A p.R352Q Chr.16: 56401763-56401763 on Build GRCh38
5693445 c.985G>A p.A329T Chr.16: 56401833-56401833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200259295 Chr.16: 56401773-56401773 on Build GRCh38
rs557288376 Chr.16: 56401873-56401873 on Build GRCh38
rs375492835 Chr.16: 56401835-56401835 on Build GRCh38
rs1016546528 Chr.16: 56401744-56401744 on Build GRCh38
rs150798359 Chr.16: 56401760-56401760 on Build GRCh38
rs371383151 Chr.16: 56401686-56401686 on Build GRCh38
rs755466258 Chr.16: 56401861-56401861 on Build GRCh38
rs774911930 Chr.16: 56401805-56401805 on Build GRCh38
rs151301190 Chr.16: 56401780-56401780 on Build GRCh38
rs764024142 Chr.16: 56401818-56401818 on Build GRCh38
rs750631855 Chr.16: 56401853-56401853 on Build GRCh38
rs369433667 Chr.16: 56401840-56401840 on Build GRCh38
rs116895221 Chr.16: 56401890-56401890 on Build GRCh38
rs751299795 Chr.16: 56401824-56401824 on Build GRCh38
rs758030819 Chr.16: 56401765-56401765 on Build GRCh38
rs778038146 Chr.16: 56401876-56401876 on Build GRCh38
rs775934391 Chr.16: 56401684-56401684 on Build GRCh38
rs763828151 Chr.16: 56401704-56401704 on Build GRCh38
rs889056775 Chr.16: 56401707-56401707 on Build GRCh38
rs780042504 Chr.16: 56401764-56401764 on Build GRCh38
rs202121611 Chr.16: 56401774-56401774 on Build GRCh38
rs368978217 Chr.16: 56401709-56401709 on Build GRCh38
rs761585749 Chr.16: 56401839-56401839 on Build GRCh38
rs373729775 Chr.16: 56401858-56401858 on Build GRCh38
rs375951425 Chr.16: 56401687-56401687 on Build GRCh38
rs749309414 Chr.16: 56401726-56401728 on Build GRCh38
rs756216374 Chr.16: 56401763-56401763 on Build GRCh38
rs1051675438 Chr.16: 56401693-56401693 on Build GRCh38
rs372095095 Chr.16: 56401755-56401755 on Build GRCh38
rs769305833 Chr.16: 56401766-56401766 on Build GRCh38
rs769316964 Chr.16: 56401800-56401800 on Build GRCh38
rs1007391937 Chr.16: 56401710-56401710 on Build GRCh38
rs759176795 Chr.16: 56401786-56401786 on Build GRCh38
rs779573010 Chr.16: 56401863-56401863 on Build GRCh38
rs368093883 Chr.16: 56401685-56401685 on Build GRCh38
rs902877141 Chr.16: 56401676-56401676 on Build GRCh38
rs745512538 Chr.16: 56401886-56401886 on Build GRCh38
rs112221607 Chr.16: 56401683-56401683 on Build GRCh38
rs754486307 Chr.16: 56401864-56401864 on Build GRCh38
rs751387600 Chr.16: 56401705-56401705 on Build GRCh38

View Full Product Details