Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11826115-11826350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDFT1
Entrez Gene ID
2222
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
DGPT, ERG9, SQS, SS
UniGene
Hs.593928
Gene Chromosome Location
Chr.8: 11795573-11839309 on Build GRCh38
Transcripts
NM_001287742.1 NM_001287743.1 NM_001287744.1 NM_001287745.1 NM_001287747.1 NM_001287748.1 NM_001287749.1 NM_001287750.1 NM_001287751.1 NM_001287756.1 NM_004462.4 XM_017013217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1095298 c.690C>A p.F230L Chr.8: 11826203-11826203 on Build GRCh38
3164733 c.614A>G p.N205S Chr.8: 11826127-11826127 on Build GRCh38
3644499 c.669C>T p.D223D Chr.8: 11826182-11826182 on Build GRCh38
5674625 c.678A>G p.G226G Chr.8: 11826191-11826191 on Build GRCh38
6031353 c.613A>G p.N205D Chr.8: 11826126-11826126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748990129 Chr.8: 11826253-11826253 on Build GRCh38
rs761055565 Chr.8: 11826137-11826137 on Build GRCh38
rs777992769 Chr.8: 11826164-11826164 on Build GRCh38
rs973798837 Chr.8: 11826237-11826237 on Build GRCh38
rs774017324 Chr.8: 11826262-11826262 on Build GRCh38
rs376692522 Chr.8: 11826195-11826195 on Build GRCh38
rs777843940 Chr.8: 11826223-11826224 on Build GRCh38
rs768195967 Chr.8: 11826191-11826191 on Build GRCh38
rs781132495 Chr.8: 11826174-11826174 on Build GRCh38
rs550011564 Chr.8: 11826319-11826319 on Build GRCh38
rs982617696 Chr.8: 11826227-11826227 on Build GRCh38
rs775185192 Chr.8: 11826185-11826185 on Build GRCh38
rs758659229 Chr.8: 11826220-11826220 on Build GRCh38
rs915720747 Chr.8: 11826210-11826210 on Build GRCh38
rs113054717 Chr.8: 11826291-11826291 on Build GRCh38
rs755329339 Chr.8: 11826143-11826143 on Build GRCh38
rs904011 Chr.8: 11826144-11826144 on Build GRCh38
rs765684285 Chr.8: 11826215-11826215 on Build GRCh38
rs764279301 Chr.8: 11826223-11826223 on Build GRCh38
rs374886414 Chr.8: 11826317-11826317 on Build GRCh38
rs762592981 Chr.8: 11826187-11826187 on Build GRCh38
rs747003322 Chr.8: 11826170-11826170 on Build GRCh38
rs762161668 Chr.8: 11826235-11826235 on Build GRCh38
rs766827610 Chr.8: 11826198-11826198 on Build GRCh38
rs186477627 Chr.8: 11826252-11826252 on Build GRCh38
rs115409999 Chr.8: 11826233-11826233 on Build GRCh38
rs747617278 Chr.8: 11826263-11826263 on Build GRCh38
rs761339090 Chr.8: 11826196-11826196 on Build GRCh38
rs892618524 Chr.8: 11826303-11826303 on Build GRCh38
rs752909843 Chr.8: 11826147-11826147 on Build GRCh38
rs769523244 Chr.8: 11826183-11826183 on Build GRCh38
rs912170889 Chr.8: 11826280-11826280 on Build GRCh38
rs2294136 Chr.8: 11826309-11826309 on Build GRCh38
rs751009166 Chr.8: 11826136-11826136 on Build GRCh38
rs373391164 Chr.8: 11826239-11826239 on Build GRCh38
rs139729511 Chr.8: 11826172-11826172 on Build GRCh38
rs373557843 Chr.8: 11826193-11826193 on Build GRCh38
rs770199595 Chr.8: 11826205-11826205 on Build GRCh38
rs766816695 Chr.8: 11826138-11826138 on Build GRCh38
rs753034680 Chr.8: 11826216-11826216 on Build GRCh38
rs369627462 Chr.8: 11826189-11826189 on Build GRCh38
rs745807159 Chr.8: 11826238-11826238 on Build GRCh38
rs754178776 Chr.8: 11826140-11826140 on Build GRCh38
rs375806521 Chr.8: 11826256-11826256 on Build GRCh38
rs144388940 Chr.8: 11826200-11826200 on Build GRCh38
rs143361983 Chr.8: 11826285-11826285 on Build GRCh38
rs567389018 Chr.8: 11826295-11826295 on Build GRCh38
rs751611810 Chr.8: 11826228-11826228 on Build GRCh38
rs751873505 Chr.8: 11826156-11826156 on Build GRCh38
rs144787232 Chr.8: 11826161-11826161 on Build GRCh38

View Full Product Details