Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137113365-137113594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP7
Entrez Gene ID
29952
Gene Name
dipeptidyl peptidase 7
Gene Aliases
DPP2, DPPII, QPP
UniGene
Hs.37916
Gene Chromosome Location
Chr.9: 137110540-137117980 on Build GRCh38
Transcripts
NM_013379.2 XM_005266075.3 XM_006717083.3 XM_011518599.2 XM_011518600.1 XM_017014651.1 XM_017014652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116555 c.578C>T p.A193V Chr.9: 137113404-137113404 on Build GRCh38
3088671 c.550G>A p.A184T Chr.9: 137113432-137113432 on Build GRCh38
1472157 c.529C>T p.H177Y Chr.9: 137113453-137113453 on Build GRCh38
6567042 c.537G>C p.V179V Chr.9: 137113445-137113445 on Build GRCh38
1107043 c.545C>T p.A182V Chr.9: 137113437-137113437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187272934 Chr.9: 137113544-137113544 on Build GRCh38
rs751246663 Chr.9: 137113383-137113383 on Build GRCh38
rs761417013 Chr.9: 137113434-137113434 on Build GRCh38
rs757478365 Chr.9: 137113453-137113453 on Build GRCh38
rs755541979 Chr.9: 137113569-137113569 on Build GRCh38
rs756303203 Chr.9: 137113557-137113557 on Build GRCh38
rs781489638 Chr.9: 137113457-137113457 on Build GRCh38
rs776801803 Chr.9: 137113454-137113455 on Build GRCh38
rs763370350 Chr.9: 137113483-137113484 on Build GRCh38
rs773795253 Chr.9: 137113490-137113490 on Build GRCh38
rs763413395 Chr.9: 137113428-137113428 on Build GRCh38
rs780433774 Chr.9: 137113563-137113563 on Build GRCh38
rs539821144 Chr.9: 137113519-137113519 on Build GRCh38
rs767453262 Chr.9: 137113540-137113540 on Build GRCh38
rs750197563 Chr.9: 137113400-137113400 on Build GRCh38
rs11547765 Chr.9: 137113440-137113440 on Build GRCh38
rs772924500 Chr.9: 137113507-137113507 on Build GRCh38
rs766635762 Chr.9: 137113452-137113453 on Build GRCh38
rs768179786 Chr.9: 137113431-137113431 on Build GRCh38
rs903028247 Chr.9: 137113537-137113537 on Build GRCh38
rs997218534 Chr.9: 137113545-137113545 on Build GRCh38
rs762647549 Chr.9: 137113436-137113436 on Build GRCh38
rs548409700 Chr.9: 137113421-137113421 on Build GRCh38
rs749644713 Chr.9: 137113564-137113564 on Build GRCh38
rs138986800 Chr.9: 137113466-137113466 on Build GRCh38
rs145102569 Chr.9: 137113460-137113460 on Build GRCh38
rs766301230 Chr.9: 137113509-137113509 on Build GRCh38
rs764655286 Chr.9: 137113560-137113561 on Build GRCh38
rs568219567 Chr.9: 137113437-137113437 on Build GRCh38
rs189319790 Chr.9: 137113463-137113463 on Build GRCh38
rs1033553220 Chr.9: 137113553-137113553 on Build GRCh38
rs895705002 Chr.9: 137113512-137113512 on Build GRCh38
rs1055700532 Chr.9: 137113483-137113483 on Build GRCh38
rs752759966 Chr.9: 137113470-137113473 on Build GRCh38
rs759713563 Chr.9: 137113457-137113460 on Build GRCh38
rs769116480 Chr.9: 137113479-137113479 on Build GRCh38
rs765253490 Chr.9: 137113521-137113521 on Build GRCh38
rs773832810 Chr.9: 137113432-137113432 on Build GRCh38
rs768405690 Chr.9: 137113443-137113443 on Build GRCh38
rs771429105 Chr.9: 137113501-137113501 on Build GRCh38
rs760207526 Chr.9: 137113508-137113508 on Build GRCh38
rs148168517 Chr.9: 137113417-137113417 on Build GRCh38
rs780123657 Chr.9: 137113405-137113405 on Build GRCh38
rs1014196954 Chr.9: 137113527-137113528 on Build GRCh38
rs760529556 Chr.9: 137113442-137113442 on Build GRCh38
rs758554118 Chr.9: 137113418-137113418 on Build GRCh38
rs752518146 Chr.9: 137113527-137113527 on Build GRCh38
rs747216596 Chr.9: 137113419-137113419 on Build GRCh38
rs774022152 Chr.9: 137113430-137113452 on Build GRCh38
rs965714858 Chr.9: 137113394-137113394 on Build GRCh38
rs767269001 Chr.9: 137113397-137113397 on Build GRCh38
rs761666033 Chr.9: 137113438-137113438 on Build GRCh38
rs776545944 Chr.9: 137113515-137113515 on Build GRCh38
rs1020640314 Chr.9: 137113548-137113548 on Build GRCh38
rs770087794 Chr.9: 137113423-137113423 on Build GRCh38
rs367853938 Chr.9: 137113422-137113422 on Build GRCh38
rs944863567 Chr.9: 137113427-137113427 on Build GRCh38
rs748539155 Chr.9: 137113574-137113574 on Build GRCh38
rs765323580 Chr.9: 137113460-137113507 on Build GRCh38
rs775982764 Chr.9: 137113424-137113424 on Build GRCh38
rs376369249 Chr.9: 137113472-137113472 on Build GRCh38
rs370348395 Chr.9: 137113384-137113384 on Build GRCh38
rs897277052 Chr.9: 137113456-137113456 on Build GRCh38
rs779491640 Chr.9: 137113573-137113573 on Build GRCh38
rs778047268 Chr.9: 137113448-137113448 on Build GRCh38
rs141922159 Chr.9: 137113451-137113451 on Build GRCh38
rs756069157 Chr.9: 137113401-137113401 on Build GRCh38
rs747684915 Chr.9: 137113491-137113491 on Build GRCh38
rs757362097 Chr.9: 137113529-137113529 on Build GRCh38
rs754913122 Chr.9: 137113445-137113445 on Build GRCh38

View Full Product Details