Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236205053-236205322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR137B
Entrez Gene ID
7107
Gene Name
G protein-coupled receptor 137B
Gene Aliases
TM7SF1
UniGene
Hs.498160
Gene Chromosome Location
Chr.1: 236142502-236211059 on Build GRCh38
Transcripts
NM_003272.3 XM_017002209.1 XM_017002210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4926593 c.997T>A p.F333I Chr.1: 236205156-236205156 on Build GRCh38
261144 c.1017C>T p.F339F Chr.1: 236205176-236205176 on Build GRCh38
6673727 c.967-7delT p.? Chr.1: 236205119-236205119 on Build GRCh38
4520821 c.1079G>A p.G360E Chr.1: 236205238-236205238 on Build GRCh38
1601948 c.1030C>T p.R344* Chr.1: 236205189-236205189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781248307 Chr.1: 236205216-236205216 on Build GRCh38
rs987122539 Chr.1: 236205197-236205197 on Build GRCh38
rs754161762 Chr.1: 236205274-236205274 on Build GRCh38
rs1001262491 Chr.1: 236205084-236205084 on Build GRCh38
rs773608709 Chr.1: 236205290-236205304 on Build GRCh38
rs971479422 Chr.1: 236205082-236205082 on Build GRCh38
rs775952091 Chr.1: 236205260-236205260 on Build GRCh38
rs138218694 Chr.1: 236205210-236205210 on Build GRCh38
rs565202568 Chr.1: 236205252-236205253 on Build GRCh38
rs769754962 Chr.1: 236205133-236205133 on Build GRCh38
rs191574434 Chr.1: 236205189-236205189 on Build GRCh38
rs771127296 Chr.1: 236205246-236205246 on Build GRCh38
rs904686485 Chr.1: 236205271-236205271 on Build GRCh38
rs751591125 Chr.1: 236205160-236205160 on Build GRCh38
rs1038698358 Chr.1: 236205225-236205225 on Build GRCh38
rs764359350 Chr.1: 236205270-236205270 on Build GRCh38
rs989969087 Chr.1: 236205150-236205151 on Build GRCh38
rs750027184 Chr.1: 236205190-236205190 on Build GRCh38
rs997089893 Chr.1: 236205231-236205231 on Build GRCh38
rs879104215 Chr.1: 236205100-236205100 on Build GRCh38
rs371587043 Chr.1: 236205262-236205262 on Build GRCh38
rs372483736 Chr.1: 236205290-236205290 on Build GRCh38
rs779573930 Chr.1: 236205206-236205206 on Build GRCh38
rs749041958 Chr.1: 236205244-236205244 on Build GRCh38
rs397840147 Chr.1: 236205092-236205093 on Build GRCh38
rs774459252 Chr.1: 236205247-236205247 on Build GRCh38
rs145077949 Chr.1: 236205226-236205226 on Build GRCh38
rs3215798 Chr.1: 236205091-236205092 on Build GRCh38
rs781585840 Chr.1: 236205173-236205176 on Build GRCh38
rs751214911 Chr.1: 236205294-236205294 on Build GRCh38
rs757572633 Chr.1: 236205279-236205279 on Build GRCh38
rs773126810 Chr.1: 236205232-236205232 on Build GRCh38
rs998983141 Chr.1: 236205283-236205283 on Build GRCh38
rs766366112 Chr.1: 236205158-236205158 on Build GRCh38
rs746483606 Chr.1: 236205209-236205209 on Build GRCh38
rs756437891 Chr.1: 236205182-236205182 on Build GRCh38
rs763295179 Chr.1: 236205156-236205156 on Build GRCh38
rs761664727 Chr.1: 236205096-236205096 on Build GRCh38
rs767623198 Chr.1: 236205256-236205256 on Build GRCh38
rs759572276 Chr.1: 236205251-236205251 on Build GRCh38
rs527975997 Chr.1: 236205153-236205153 on Build GRCh38
rs201754513 Chr.1: 236205180-236205180 on Build GRCh38
rs373821559 Chr.1: 236205176-236205176 on Build GRCh38
rs929092444 Chr.1: 236205144-236205144 on Build GRCh38
rs776798717 Chr.1: 236205111-236205114 on Build GRCh38
rs758051106 Chr.1: 236205148-236205148 on Build GRCh38
rs776735600 Chr.1: 236205093-236205093 on Build GRCh38

View Full Product Details