Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66822311-66822536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf44-SGK3
Entrez Gene ID
100533105
Gene Name
C8orf44-SGK3 readthrough
Gene Aliases
-
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66667552-66862022 on Build GRCh38
Transcripts
NM_001204173.1 NM_001033578.2 NM_013257.4 NM_170709.2

Gene Symbol
SGK3
Entrez Gene ID
23678
Gene Name
serum/glucocorticoid regulated kinase family member 3
Gene Aliases
CISK, SGK2, SGKL
UniGene
Hs.613417
Gene Chromosome Location
Chr.8: 66712418-66862022 on Build GRCh38
Transcripts
NM_001204173.1 NM_001033578.2 NM_013257.4 NM_170709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400701 c.417+2T>C p.? Chr.8: 66822461-66822461 on Build GRCh38
1457884 c.209C>A p.S70Y Chr.8: 66822452-66822452 on Build GRCh38
6113574 c.138C>T p.V46V Chr.8: 66822381-66822381 on Build GRCh38
5932303 c.129-7T>C p.? Chr.8: 66822365-66822365 on Build GRCh38
400702 c.216+2T>C p.? Chr.8: 66822461-66822461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749670901 Chr.8: 66822475-66822475 on Build GRCh38
rs376914097 Chr.8: 66822340-66822340 on Build GRCh38
rs781646437 Chr.8: 66822426-66822426 on Build GRCh38
rs753691709 Chr.8: 66822386-66822386 on Build GRCh38
rs926392545 Chr.8: 66822348-66822348 on Build GRCh38
rs950238767 Chr.8: 66822396-66822396 on Build GRCh38
rs374276019 Chr.8: 66822495-66822495 on Build GRCh38
rs755143437 Chr.8: 66822421-66822421 on Build GRCh38
rs990517924 Chr.8: 66822379-66822379 on Build GRCh38
rs770314606 Chr.8: 66822467-66822467 on Build GRCh38
rs574203589 Chr.8: 66822413-66822413 on Build GRCh38
rs763941506 Chr.8: 66822385-66822385 on Build GRCh38
rs201134569 Chr.8: 66822383-66822383 on Build GRCh38
rs770488800 Chr.8: 66822492-66822492 on Build GRCh38
rs934872499 Chr.8: 66822507-66822507 on Build GRCh38
rs747055036 Chr.8: 66822403-66822403 on Build GRCh38
rs1009495474 Chr.8: 66822429-66822429 on Build GRCh38
rs371705415 Chr.8: 66822465-66822465 on Build GRCh38
rs774762060 Chr.8: 66822478-66822478 on Build GRCh38
rs748689772 Chr.8: 66822466-66822466 on Build GRCh38
rs890597454 Chr.8: 66822427-66822427 on Build GRCh38
rs1053413373 Chr.8: 66822376-66822376 on Build GRCh38
rs752763955 Chr.8: 66822494-66822495 on Build GRCh38
rs184264216 Chr.8: 66822440-66822440 on Build GRCh38
rs780326958 Chr.8: 66822400-66822400 on Build GRCh38
rs767552397 Chr.8: 66822365-66822365 on Build GRCh38
rs766231182 Chr.8: 66822488-66822489 on Build GRCh38
rs189621437 Chr.8: 66822468-66822468 on Build GRCh38
rs752734301 Chr.8: 66822373-66822373 on Build GRCh38
rs758491010 Chr.8: 66822399-66822399 on Build GRCh38
rs199750865 Chr.8: 66822358-66822358 on Build GRCh38
rs981797948 Chr.8: 66822341-66822341 on Build GRCh38

View Full Product Details