Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54615931-54616198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_176782.2 XM_006710599.3

Gene Symbol
FAM151A
Entrez Gene ID
338094
Gene Name
family with sequence similarity 151 member A
Gene Aliases
C1orf179
UniGene
Hs.234786
Gene Chromosome Location
Chr.1: 54609177-54623527 on Build GRCh38
Transcripts
NM_015547.3 NM_176782.2 XM_006710599.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623055 c.345C>G p.H115Q Chr.1: 54616090-54616090 on Build GRCh38
6009291 c.319G>A p.E107K Chr.1: 54616116-54616116 on Build GRCh38
4577452 c.390C>T p.D130D Chr.1: 54616045-54616045 on Build GRCh38
1748516 c.333C>G p.P111P Chr.1: 54616102-54616102 on Build GRCh38
1343509 c.351_352insC p.T118fs*5 Chr.1: 54616083-54616084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777059546 Chr.1: 54616042-54616042 on Build GRCh38
rs779759267 Chr.1: 54616008-54616008 on Build GRCh38
rs759428311 Chr.1: 54616146-54616146 on Build GRCh38
rs771500117 Chr.1: 54616125-54616125 on Build GRCh38
rs765149719 Chr.1: 54616147-54616147 on Build GRCh38
rs766409875 Chr.1: 54616131-54616131 on Build GRCh38
rs773943398 Chr.1: 54616106-54616106 on Build GRCh38
rs1041995144 Chr.1: 54615980-54615980 on Build GRCh38
rs201533192 Chr.1: 54616167-54616167 on Build GRCh38
rs758503125 Chr.1: 54616086-54616086 on Build GRCh38
rs200866369 Chr.1: 54616089-54616089 on Build GRCh38
rs772679617 Chr.1: 54616018-54616018 on Build GRCh38
rs987917732 Chr.1: 54615990-54615990 on Build GRCh38
rs918100033 Chr.1: 54616087-54616087 on Build GRCh38
rs776873902 Chr.1: 54616175-54616175 on Build GRCh38
rs200435280 Chr.1: 54615993-54615993 on Build GRCh38
rs773523016 Chr.1: 54616119-54616119 on Build GRCh38
rs914713537 Chr.1: 54615954-54615954 on Build GRCh38
rs965838626 Chr.1: 54615975-54615975 on Build GRCh38
rs750196655 Chr.1: 54615983-54615983 on Build GRCh38
rs755812938 Chr.1: 54615987-54615987 on Build GRCh38
rs1018806264 Chr.1: 54615970-54615970 on Build GRCh38
rs748837133 Chr.1: 54616044-54616044 on Build GRCh38
rs202087836 Chr.1: 54616128-54616128 on Build GRCh38
rs767482184 Chr.1: 54615973-54615973 on Build GRCh38
rs199649539 Chr.1: 54616118-54616118 on Build GRCh38
rs371885641 Chr.1: 54616090-54616091 on Build GRCh38
rs201220765 Chr.1: 54616115-54616115 on Build GRCh38
rs201399565 Chr.1: 54615992-54615992 on Build GRCh38
rs748308425 Chr.1: 54616014-54616014 on Build GRCh38
rs772698420 Chr.1: 54616090-54616090 on Build GRCh38
rs142814457 Chr.1: 54616168-54616168 on Build GRCh38
rs143281849 Chr.1: 54616100-54616100 on Build GRCh38
rs17851843 Chr.1: 54616079-54616079 on Build GRCh38
rs747541842 Chr.1: 54616026-54616026 on Build GRCh38
rs776262199 Chr.1: 54616135-54616135 on Build GRCh38
rs139479416 Chr.1: 54616099-54616099 on Build GRCh38
rs889764954 Chr.1: 54616158-54616158 on Build GRCh38
rs540583125 Chr.1: 54616045-54616045 on Build GRCh38
rs761118154 Chr.1: 54616139-54616139 on Build GRCh38
rs766027088 Chr.1: 54616080-54616080 on Build GRCh38
rs938268268 Chr.1: 54616081-54616081 on Build GRCh38
rs77612989 Chr.1: 54616127-54616127 on Build GRCh38
rs1039717077 Chr.1: 54616178-54616178 on Build GRCh38
rs761236004 Chr.1: 54616001-54616015 on Build GRCh38
rs761361008 Chr.1: 54616054-54616054 on Build GRCh38
rs767036550 Chr.1: 54616067-54616067 on Build GRCh38
rs748730950 Chr.1: 54616063-54616063 on Build GRCh38
rs753726688 Chr.1: 54616084-54616084 on Build GRCh38
rs752805978 Chr.1: 54616085-54616085 on Build GRCh38
rs749953492 Chr.1: 54616084-54616084 on Build GRCh38
rs940182473 Chr.1: 54616124-54616124 on Build GRCh38
rs1007441965 Chr.1: 54615962-54615962 on Build GRCh38
rs773090202 Chr.1: 54616072-54616072 on Build GRCh38
rs764550087 Chr.1: 54616083-54616084 on Build GRCh38
rs771161344 Chr.1: 54616038-54616038 on Build GRCh38
rs779126139 Chr.1: 54615999-54615999 on Build GRCh38
rs760514214 Chr.1: 54616129-54616129 on Build GRCh38
rs778478162 Chr.1: 54616020-54616020 on Build GRCh38
rs781404698 Chr.1: 54616095-54616095 on Build GRCh38
rs926276661 Chr.1: 54616035-54616035 on Build GRCh38
rs760662505 Chr.1: 54616074-54616074 on Build GRCh38
rs773818587 Chr.1: 54616048-54616048 on Build GRCh38
rs1027504698 Chr.1: 54616053-54616053 on Build GRCh38
rs755239601 Chr.1: 54615994-54615994 on Build GRCh38

View Full Product Details