Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65974197-65974426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677199 c.1376C>A p.A459D Chr.7: 65974310-65974310 on Build GRCh38
3085623 c.1373C>G p.S458C Chr.7: 65974313-65974313 on Build GRCh38
6340246 c.1347C>A p.A449A Chr.7: 65974339-65974339 on Build GRCh38
6677193 c.1324G>A p.A442T Chr.7: 65974362-65974362 on Build GRCh38
3085627 c.1260C>T p.N420N Chr.7: 65974426-65974426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376131143 Chr.7: 65974401-65974401 on Build GRCh38
rs751894676 Chr.7: 65974351-65974351 on Build GRCh38
rs748839927 Chr.7: 65974258-65974258 on Build GRCh38
rs746796155 Chr.7: 65974285-65974285 on Build GRCh38
rs368144710 Chr.7: 65974363-65974363 on Build GRCh38
rs753483823 Chr.7: 65974362-65974362 on Build GRCh38
rs912985278 Chr.7: 65974328-65974328 on Build GRCh38
rs1026800808 Chr.7: 65974226-65974226 on Build GRCh38
rs758314777 Chr.7: 65974383-65974383 on Build GRCh38
rs768780031 Chr.7: 65974290-65974290 on Build GRCh38
rs121918180 Chr.7: 65974348-65974348 on Build GRCh38
rs761881997 Chr.7: 65974317-65974317 on Build GRCh38
rs914678759 Chr.7: 65974234-65974234 on Build GRCh38
rs769803627 Chr.7: 65974325-65974325 on Build GRCh38
rs377217843 Chr.7: 65974254-65974254 on Build GRCh38
rs768543128 Chr.7: 65974384-65974386 on Build GRCh38
rs376567545 Chr.7: 65974336-65974336 on Build GRCh38
rs375941316 Chr.7: 65974232-65974232 on Build GRCh38
rs763845326 Chr.7: 65974361-65974361 on Build GRCh38
rs199806689 Chr.7: 65974267-65974267 on Build GRCh38
rs143040697 Chr.7: 65974395-65974395 on Build GRCh38
rs113484908 Chr.7: 65974270-65974270 on Build GRCh38
rs369767887 Chr.7: 65974389-65974389 on Build GRCh38
rs118066970 Chr.7: 65974327-65974327 on Build GRCh38
rs987261453 Chr.7: 65974233-65974233 on Build GRCh38
rs766849255 Chr.7: 65974397-65974397 on Build GRCh38
rs934656347 Chr.7: 65974231-65974231 on Build GRCh38
rs911843328 Chr.7: 65974244-65974244 on Build GRCh38
rs1002438938 Chr.7: 65974392-65974392 on Build GRCh38
rs35957142 Chr.7: 65974366-65974367 on Build GRCh38
rs772040985 Chr.7: 65974272-65974272 on Build GRCh38
rs768428238 Chr.7: 65974398-65974398 on Build GRCh38
rs139776224 Chr.7: 65974357-65974357 on Build GRCh38
rs776791543 Chr.7: 65974303-65974303 on Build GRCh38
rs368233882 Chr.7: 65974271-65974271 on Build GRCh38
rs777436987 Chr.7: 65974257-65974257 on Build GRCh38
rs778631803 Chr.7: 65974373-65974373 on Build GRCh38
rs749917538 Chr.7: 65974382-65974382 on Build GRCh38

View Full Product Details