Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29145051-29145306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP4
Entrez Gene ID
79745
Gene Name
CAP-Gly domain containing linker protein family member 4
Gene Aliases
RSNL2
UniGene
Hs.122927
Gene Chromosome Location
Chr.2: 29097676-29183813 on Build GRCh38
Transcripts
NM_001287527.1 NM_001287528.1 NM_024692.5 NR_109844.1 XM_005264562.2 XM_005264563.1 XM_006712104.2 XM_006712105.3 XM_011533109.1 XM_011533110.1 XM_017004958.1 XM_017004959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5829457 c.922G>A p.A308T Chr.2: 29145268-29145268 on Build GRCh38
1631968 c.901T>C p.F301L Chr.2: 29145247-29145247 on Build GRCh38
1407650 c.922G>A p.A308T Chr.2: 29145268-29145268 on Build GRCh38
4672840 c.891T>C p.G297G Chr.2: 29145237-29145237 on Build GRCh38
1019997 c.926G>A p.S309N Chr.2: 29145272-29145272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055437870 Chr.2: 29145178-29145178 on Build GRCh38
rs749638451 Chr.2: 29145201-29145201 on Build GRCh38
rs765873203 Chr.2: 29145278-29145278 on Build GRCh38
rs753746547 Chr.2: 29145239-29145239 on Build GRCh38
rs374159869 Chr.2: 29145240-29145240 on Build GRCh38
rs1053400366 Chr.2: 29145098-29145098 on Build GRCh38
rs554117829 Chr.2: 29145092-29145092 on Build GRCh38
rs370186094 Chr.2: 29145212-29145212 on Build GRCh38
rs1004109994 Chr.2: 29145267-29145267 on Build GRCh38
rs935754406 Chr.2: 29145093-29145093 on Build GRCh38
rs978092039 Chr.2: 29145133-29145133 on Build GRCh38
rs575913843 Chr.2: 29145210-29145210 on Build GRCh38
rs765309019 Chr.2: 29145245-29145245 on Build GRCh38
rs967644488 Chr.2: 29145127-29145127 on Build GRCh38
rs547770504 Chr.2: 29145152-29145152 on Build GRCh38
rs915654278 Chr.2: 29145195-29145195 on Build GRCh38
rs891606160 Chr.2: 29145131-29145131 on Build GRCh38
rs914765516 Chr.2: 29145091-29145091 on Build GRCh38
rs998538841 Chr.2: 29145151-29145151 on Build GRCh38
rs757869190 Chr.2: 29145275-29145275 on Build GRCh38
rs778045804 Chr.2: 29145199-29145199 on Build GRCh38
rs546292788 Chr.2: 29145220-29145220 on Build GRCh38
rs889802156 Chr.2: 29145167-29145167 on Build GRCh38
rs928299257 Chr.2: 29145164-29145164 on Build GRCh38
rs760881993 Chr.2: 29145222-29145222 on Build GRCh38
rs776649793 Chr.2: 29145081-29145081 on Build GRCh38
rs1026262821 Chr.2: 29145221-29145221 on Build GRCh38
rs750368640 Chr.2: 29145264-29145264 on Build GRCh38
rs751178030 Chr.2: 29145281-29145281 on Build GRCh38
rs770088241 Chr.2: 29145183-29145183 on Build GRCh38
rs992840808 Chr.2: 29145259-29145259 on Build GRCh38
rs772137138 Chr.2: 29145219-29145219 on Build GRCh38
rs763807915 Chr.2: 29145224-29145224 on Build GRCh38
rs771492166 Chr.2: 29145203-29145203 on Build GRCh38
rs565832402 Chr.2: 29145123-29145123 on Build GRCh38

View Full Product Details