Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110645858-110646127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
UniGene
Hs.508725
Gene Chromosome Location
Chr.13: 110641410-110713527 on Build GRCh38
Transcripts
NM_024537.3 XM_006719953.2 XM_011521116.1 XM_017020741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6200017 c.1199C>G p.S400C Chr.13: 110646085-110646085 on Build GRCh38
3753581 c.1239T>C p.D413D Chr.13: 110646045-110646045 on Build GRCh38
945388 c.1307C>T p.A436V Chr.13: 110645977-110645977 on Build GRCh38
3813402 c.1209G>C p.K403N Chr.13: 110646075-110646075 on Build GRCh38
4524715 c.1289G>A p.G430E Chr.13: 110645995-110645995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377292484 Chr.13: 110646107-110646107 on Build GRCh38
rs758399378 Chr.13: 110645920-110645920 on Build GRCh38
rs780804270 Chr.13: 110645955-110645956 on Build GRCh38
rs436462 Chr.13: 110646045-110646045 on Build GRCh38
rs374773952 Chr.13: 110645915-110645915 on Build GRCh38
rs775014066 Chr.13: 110645935-110645935 on Build GRCh38
rs368099040 Chr.13: 110646056-110646056 on Build GRCh38
rs946100593 Chr.13: 110646026-110646026 on Build GRCh38
rs556643017 Chr.13: 110646078-110646078 on Build GRCh38
rs766810393 Chr.13: 110645934-110645934 on Build GRCh38
rs143945861 Chr.13: 110645997-110645997 on Build GRCh38
rs752221052 Chr.13: 110646067-110646067 on Build GRCh38
rs770340849 Chr.13: 110645927-110645927 on Build GRCh38
rs1020669863 Chr.13: 110645891-110645891 on Build GRCh38
rs369682310 Chr.13: 110645950-110645950 on Build GRCh38
rs138825549 Chr.13: 110645887-110645887 on Build GRCh38
rs920168493 Chr.13: 110645962-110645962 on Build GRCh38
rs762700157 Chr.13: 110646077-110646077 on Build GRCh38
rs750186107 Chr.13: 110645974-110645974 on Build GRCh38
rs1032230756 Chr.13: 110645916-110645916 on Build GRCh38
rs946151296 Chr.13: 110645890-110645890 on Build GRCh38
rs202162305 Chr.13: 110646007-110646007 on Build GRCh38
rs141349632 Chr.13: 110645976-110645976 on Build GRCh38
rs923772737 Chr.13: 110646054-110646054 on Build GRCh38
rs770146415 Chr.13: 110646009-110646009 on Build GRCh38
rs193086198 Chr.13: 110646060-110646060 on Build GRCh38
rs749638271 Chr.13: 110646019-110646019 on Build GRCh38
rs748447663 Chr.13: 110646002-110646002 on Build GRCh38
rs185831746 Chr.13: 110646103-110646103 on Build GRCh38
rs753537619 Chr.13: 110645946-110645946 on Build GRCh38
rs376207512 Chr.13: 110645941-110645941 on Build GRCh38
rs745872506 Chr.13: 110645919-110645919 on Build GRCh38
rs146878400 Chr.13: 110646025-110646025 on Build GRCh38
rs1008383099 Chr.13: 110645879-110645879 on Build GRCh38
rs900394244 Chr.13: 110646082-110646082 on Build GRCh38
rs757028283 Chr.13: 110645956-110645956 on Build GRCh38
rs144612077 Chr.13: 110646066-110646066 on Build GRCh38
rs548716694 Chr.13: 110645992-110645992 on Build GRCh38
rs777038198 Chr.13: 110645925-110645925 on Build GRCh38
rs764879319 Chr.13: 110646061-110646061 on Build GRCh38
rs774758878 Chr.13: 110646027-110646027 on Build GRCh38
rs747539005 Chr.13: 110646087-110646087 on Build GRCh38
rs766539038 Chr.13: 110646034-110646034 on Build GRCh38
rs374377972 Chr.13: 110646068-110646068 on Build GRCh38
rs568526596 Chr.13: 110646097-110646097 on Build GRCh38
rs202176952 Chr.13: 110646089-110646089 on Build GRCh38
rs116312064 Chr.13: 110645940-110645940 on Build GRCh38
rs890904519 Chr.13: 110645878-110645878 on Build GRCh38
rs141996297 Chr.13: 110645957-110645957 on Build GRCh38
rs773655449 Chr.13: 110646018-110646018 on Build GRCh38
rs755877488 Chr.13: 110646105-110646106 on Build GRCh38
rs372760421 Chr.13: 110646093-110646093 on Build GRCh38
rs751587477 Chr.13: 110646035-110646035 on Build GRCh38
rs773467148 Chr.13: 110645979-110645979 on Build GRCh38
rs900154460 Chr.13: 110645924-110645924 on Build GRCh38
rs776257494 Chr.13: 110646057-110646057 on Build GRCh38
rs755161479 Chr.13: 110645993-110645993 on Build GRCh38
rs763451673 Chr.13: 110645929-110645929 on Build GRCh38
rs530176962 Chr.13: 110645977-110645977 on Build GRCh38
rs114840063 Chr.13: 110645943-110645943 on Build GRCh38
rs927563113 Chr.13: 110645997-110645999 on Build GRCh38
rs780112388 Chr.13: 110645921-110645921 on Build GRCh38
rs768885849 Chr.13: 110645922-110645922 on Build GRCh38
rs113033050 Chr.13: 110645996-110645996 on Build GRCh38
rs966326753 Chr.13: 110645903-110645903 on Build GRCh38

View Full Product Details