Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162855573-162855846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC190
Entrez Gene ID
339512
Gene Name
coiled-coil domain containing 190
Gene Aliases
C1orf110
UniGene
Hs.407631
Gene Chromosome Location
Chr.1: 162851291-162868918 on Build GRCh38
Transcripts
NM_178550.4 XM_005245125.3 XM_006711289.3 XM_011509477.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2088317 c.314G>T p.R105I Chr.1: 162855632-162855632 on Build GRCh38
4955627 c.260G>A p.G87E Chr.1: 162855671-162855671 on Build GRCh38
228279 c.271C>G p.Q91E Chr.1: 162855675-162855675 on Build GRCh38
4224598 c.299G>T p.R100I Chr.1: 162855632-162855632 on Build GRCh38
4955626 c.275G>A p.G92E Chr.1: 162855671-162855671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747998813 Chr.1: 162855688-162855688 on Build GRCh38
rs758458325 Chr.1: 162855663-162855663 on Build GRCh38
rs572837238 Chr.1: 162855608-162855608 on Build GRCh38
rs759039651 Chr.1: 162855614-162855615 on Build GRCh38
rs751670608 Chr.1: 162855764-162855764 on Build GRCh38
rs766885894 Chr.1: 162855714-162855714 on Build GRCh38
rs754951245 Chr.1: 162855678-162855678 on Build GRCh38
rs768289439 Chr.1: 162855602-162855602 on Build GRCh38
rs766995543 Chr.1: 162855695-162855697 on Build GRCh38
rs915474849 Chr.1: 162855706-162855706 on Build GRCh38
rs377737928 Chr.1: 162855726-162855726 on Build GRCh38
rs773940880 Chr.1: 162855704-162855704 on Build GRCh38
rs762512371 Chr.1: 162855619-162855619 on Build GRCh38
rs536907515 Chr.1: 162855722-162855722 on Build GRCh38
rs776523013 Chr.1: 162855604-162855604 on Build GRCh38
rs139941480 Chr.1: 162855700-162855700 on Build GRCh38
rs77227460 Chr.1: 162855667-162855667 on Build GRCh38
rs754853570 Chr.1: 162855768-162855768 on Build GRCh38
rs1030889671 Chr.1: 162855718-162855718 on Build GRCh38
rs965441996 Chr.1: 162855612-162855612 on Build GRCh38
rs756039250 Chr.1: 162855730-162855731 on Build GRCh38
rs772887259 Chr.1: 162855693-162855693 on Build GRCh38
rs777570231 Chr.1: 162855792-162855792 on Build GRCh38
rs919620061 Chr.1: 162855816-162855816 on Build GRCh38
rs764643155 Chr.1: 162855731-162855733 on Build GRCh38
rs369304295 Chr.1: 162855639-162855639 on Build GRCh38
rs542951405 Chr.1: 162855784-162855789 on Build GRCh38
rs375996919 Chr.1: 162855618-162855618 on Build GRCh38
rs746883277 Chr.1: 162855673-162855673 on Build GRCh38
rs147909245 Chr.1: 162855611-162855611 on Build GRCh38
rs755955596 Chr.1: 162855790-162855790 on Build GRCh38
rs896536006 Chr.1: 162855649-162855650 on Build GRCh38
rs1045127574 Chr.1: 162855658-162855658 on Build GRCh38
rs368164758 Chr.1: 162855774-162855774 on Build GRCh38
rs748855512 Chr.1: 162855805-162855805 on Build GRCh38
rs753190588 Chr.1: 162855621-162855621 on Build GRCh38
rs765622936 Chr.1: 162855656-162855656 on Build GRCh38
rs769453631 Chr.1: 162855616-162855616 on Build GRCh38
rs747910581 Chr.1: 162855776-162855776 on Build GRCh38
rs768033654 Chr.1: 162855620-162855620 on Build GRCh38
rs373634463 Chr.1: 162855646-162855646 on Build GRCh38
rs765534003 Chr.1: 162855728-162855728 on Build GRCh38
rs754258885 Chr.1: 162855648-162855648 on Build GRCh38
rs758676134 Chr.1: 162855750-162855750 on Build GRCh38
rs746798567 Chr.1: 162855601-162855601 on Build GRCh38
rs780910438 Chr.1: 162855685-162855685 on Build GRCh38
rs769427289 Chr.1: 162855692-162855692 on Build GRCh38
rs750622469 Chr.1: 162855737-162855737 on Build GRCh38
rs748771375 Chr.1: 162855694-162855694 on Build GRCh38
rs377556745 Chr.1: 162855650-162855650 on Build GRCh38
rs370258122 Chr.1: 162855763-162855763 on Build GRCh38
rs761018736 Chr.1: 162855709-162855709 on Build GRCh38
rs752668355 Chr.1: 162855729-162855731 on Build GRCh38

View Full Product Details