Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49251299-49251534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs782267196 Chr.X: 49251481-49251481 on Build GRCh38
rs782632308 Chr.X: 49251496-49251496 on Build GRCh38
rs201723322 Chr.X: 49251430-49251430 on Build GRCh38
rs145368924 Chr.X: 49251381-49251381 on Build GRCh38
rs781802008 Chr.X: 49251418-49251418 on Build GRCh38
rs782088695 Chr.X: 49251345-49251345 on Build GRCh38
rs782112916 Chr.X: 49251335-49251335 on Build GRCh38
rs782478365 Chr.X: 49251403-49251403 on Build GRCh38
rs979425659 Chr.X: 49251337-49251337 on Build GRCh38
rs782786757 Chr.X: 49251389-49251389 on Build GRCh38
rs373302430 Chr.X: 49251409-49251409 on Build GRCh38
rs781854593 Chr.X: 49251390-49251390 on Build GRCh38
rs28935477 Chr.X: 49251441-49251441 on Build GRCh38
rs369332983 Chr.X: 49251352-49251352 on Build GRCh38
rs122467170 Chr.X: 49251480-49251480 on Build GRCh38
rs925367928 Chr.X: 49251380-49251380 on Build GRCh38
rs781796588 Chr.X: 49251376-49251376 on Build GRCh38
rs370285865 Chr.X: 49251495-49251495 on Build GRCh38
rs782232479 Chr.X: 49251497-49251497 on Build GRCh38

View Full Product Details