Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45811331-45811602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID2
Entrez Gene ID
196528
Gene Name
AT-rich interaction domain 2
Gene Aliases
BAF200, p200
UniGene
Hs.317304
Gene Chromosome Location
Chr.12: 45729683-45908037 on Build GRCh38
Transcripts
NM_152641.2 XM_006719272.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6845481 c.285-1G>T p.? Chr.12: 45811417-45811417 on Build GRCh38
6915149 c.416C>T p.S139L Chr.12: 45811549-45811549 on Build GRCh38
4767428 c.310C>A p.H104N Chr.12: 45811443-45811443 on Build GRCh38
3460960 c.366T>G p.L122L Chr.12: 45811499-45811499 on Build GRCh38
6944940 c.352_367del16 p.P118fs*92 Chr.12: 45811485-45811500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778930136 Chr.12: 45811522-45811522 on Build GRCh38
rs959618294 Chr.12: 45811483-45811483 on Build GRCh38
rs778897506 Chr.12: 45811374-45811374 on Build GRCh38
rs777430761 Chr.12: 45811565-45811565 on Build GRCh38
rs1025267667 Chr.12: 45811491-45811491 on Build GRCh38
rs369360302 Chr.12: 45811394-45811394 on Build GRCh38
rs770238875 Chr.12: 45811454-45811454 on Build GRCh38
rs73292513 Chr.12: 45811472-45811472 on Build GRCh38
rs1053148714 Chr.12: 45811363-45811363 on Build GRCh38
rs1028736703 Chr.12: 45811476-45811476 on Build GRCh38
rs777169222 Chr.12: 45811408-45811409 on Build GRCh38
rs777429678 Chr.12: 45811393-45811393 on Build GRCh38
rs567746850 Chr.12: 45811433-45811433 on Build GRCh38
rs761936321 Chr.12: 45811497-45811497 on Build GRCh38
rs138168440 Chr.12: 45811484-45811484 on Build GRCh38
rs147582662 Chr.12: 45811481-45811481 on Build GRCh38
rs746631889 Chr.12: 45811400-45811400 on Build GRCh38
rs747901002 Chr.12: 45811379-45811379 on Build GRCh38
rs142032280 Chr.12: 45811503-45811503 on Build GRCh38
rs373958652 Chr.12: 45811380-45811380 on Build GRCh38
rs758330574 Chr.12: 45811563-45811563 on Build GRCh38
rs202109070 Chr.12: 45811544-45811544 on Build GRCh38
rs771857499 Chr.12: 45811385-45811385 on Build GRCh38
rs901516090 Chr.12: 45811375-45811375 on Build GRCh38
rs763234710 Chr.12: 45811469-45811469 on Build GRCh38
rs775866001 Chr.12: 45811462-45811462 on Build GRCh38
rs746714850 Chr.12: 45811573-45811573 on Build GRCh38
rs1009211953 Chr.12: 45811382-45811382 on Build GRCh38
rs754930736 Chr.12: 45811368-45811368 on Build GRCh38
rs375773959 Chr.12: 45811509-45811509 on Build GRCh38
rs140285438 Chr.12: 45811425-45811425 on Build GRCh38
rs746854058 Chr.12: 45811457-45811458 on Build GRCh38
rs770544852 Chr.12: 45811424-45811424 on Build GRCh38
rs750443564 Chr.12: 45811504-45811504 on Build GRCh38

View Full Product Details