Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37109105-37109366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK2
Entrez Gene ID
5610
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases
EIF2AK1, PKR, PPP1R83, PRKR
UniGene
Hs.131431
Gene Chromosome Location
Chr.2: 37099210-37157065 on Build GRCh38
Transcripts
NM_001135651.2 NM_001135652.2 NM_002759.3 XM_011532987.2 XM_017004503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
368735 c.1446T>G p.L482L Chr.2: 37109227-37109227 on Build GRCh38
6587479 c.1442T>G p.L481R Chr.2: 37109231-37109231 on Build GRCh38
1020691 c.1408G>T p.D470Y Chr.2: 37109265-37109265 on Build GRCh38
5633587 c.1450_1451insT p.C485fs*2 Chr.2: 37109222-37109223 on Build GRCh38
6652633 c.1408G>A p.D470N Chr.2: 37109265-37109265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752198346 Chr.2: 37109178-37109178 on Build GRCh38
rs772956630 Chr.2: 37109162-37109162 on Build GRCh38
rs139703307 Chr.2: 37109206-37109206 on Build GRCh38
rs770949145 Chr.2: 37109283-37109283 on Build GRCh38
rs201956480 Chr.2: 37109153-37109153 on Build GRCh38
rs769042439 Chr.2: 37109143-37109143 on Build GRCh38
rs762962032 Chr.2: 37109169-37109169 on Build GRCh38
rs78605520 Chr.2: 37109286-37109286 on Build GRCh38
rs779404171 Chr.2: 37109236-37109236 on Build GRCh38
rs376530608 Chr.2: 37109257-37109257 on Build GRCh38
rs1034235001 Chr.2: 37109166-37109166 on Build GRCh38
rs148773664 Chr.2: 37109272-37109272 on Build GRCh38
rs774169619 Chr.2: 37109171-37109171 on Build GRCh38
rs374591026 Chr.2: 37109232-37109232 on Build GRCh38
rs777002057 Chr.2: 37109167-37109170 on Build GRCh38
rs766041173 Chr.2: 37109170-37109170 on Build GRCh38
rs767214445 Chr.2: 37109173-37109173 on Build GRCh38
rs760394593 Chr.2: 37109319-37109319 on Build GRCh38
rs977281418 Chr.2: 37109268-37109268 on Build GRCh38
rs749142819 Chr.2: 37109279-37109279 on Build GRCh38
rs371033727 Chr.2: 37109200-37109200 on Build GRCh38
rs759165571 Chr.2: 37109172-37109172 on Build GRCh38
rs747429747 Chr.2: 37109256-37109256 on Build GRCh38
rs751013570 Chr.2: 37109298-37109299 on Build GRCh38
rs774118494 Chr.2: 37109287-37109287 on Build GRCh38
rs759392578 Chr.2: 37109288-37109288 on Build GRCh38
rs2307469 Chr.2: 37109260-37109260 on Build GRCh38
rs374043257 Chr.2: 37109196-37109196 on Build GRCh38
rs780536929 Chr.2: 37109224-37109224 on Build GRCh38
rs767300202 Chr.2: 37109296-37109296 on Build GRCh38
rs377628551 Chr.2: 37109185-37109185 on Build GRCh38
rs781631870 Chr.2: 37109258-37109258 on Build GRCh38
rs370235809 Chr.2: 37109305-37109305 on Build GRCh38
rs757758492 Chr.2: 37109226-37109228 on Build GRCh38
rs776833510 Chr.2: 37109145-37109145 on Build GRCh38
rs763470155 Chr.2: 37109328-37109328 on Build GRCh38
rs769765973 Chr.2: 37109161-37109161 on Build GRCh38
rs769706485 Chr.2: 37109269-37109269 on Build GRCh38
rs967135397 Chr.2: 37109262-37109262 on Build GRCh38

View Full Product Details