Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 760060-760325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
UniGene
Hs.279061
Gene Chromosome Location
Chr.17: 759309-782331 on Build GRCh38
Transcripts
NM_016080.3 XM_006721540.2 XM_017024711.1 XM_017024712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4904193 c.925C>T p.P309S Chr.17: 760190-760190 on Build GRCh38
4597626 c.887C>A p.A296E Chr.17: 760228-760228 on Build GRCh38
5426485 c.888A>T p.A296A Chr.17: 760227-760227 on Build GRCh38
4597625 c.842C>A p.A281E Chr.17: 760228-760228 on Build GRCh38
3521248 c.880C>T p.P294S Chr.17: 760190-760190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761777623 Chr.17: 760277-760277 on Build GRCh38
rs75027378 Chr.17: 760228-760228 on Build GRCh38
rs576834747 Chr.17: 760113-760113 on Build GRCh38
rs1045630938 Chr.17: 760262-760262 on Build GRCh38
rs770288179 Chr.17: 760234-760234 on Build GRCh38
rs769263989 Chr.17: 760124-760134 on Build GRCh38
rs185701966 Chr.17: 760099-760099 on Build GRCh38
rs780730872 Chr.17: 760171-760171 on Build GRCh38
rs895474099 Chr.17: 760107-760107 on Build GRCh38
rs1004307021 Chr.17: 760093-760093 on Build GRCh38
rs1013954506 Chr.17: 760108-760108 on Build GRCh38
rs758282527 Chr.17: 760224-760224 on Build GRCh38
rs577219986 Chr.17: 760182-760182 on Build GRCh38
rs934514210 Chr.17: 760253-760253 on Build GRCh38
rs7342834 Chr.17: 760106-760106 on Build GRCh38
rs367623782 Chr.17: 760204-760204 on Build GRCh38
rs528682390 Chr.17: 760088-760088 on Build GRCh38
rs140880281 Chr.17: 760172-760172 on Build GRCh38
rs751857074 Chr.17: 760226-760226 on Build GRCh38
rs776569387 Chr.17: 760138-760139 on Build GRCh38
rs558366243 Chr.17: 760112-760112 on Build GRCh38
rs116004401 Chr.17: 760215-760215 on Build GRCh38
rs77862410 Chr.17: 760188-760188 on Build GRCh38
rs776297488 Chr.17: 760265-760265 on Build GRCh38
rs200190271 Chr.17: 760233-760233 on Build GRCh38
rs957455719 Chr.17: 760223-760223 on Build GRCh38
rs367979387 Chr.17: 760235-760235 on Build GRCh38
rs768235571 Chr.17: 760264-760264 on Build GRCh38
rs765263827 Chr.17: 760278-760278 on Build GRCh38
rs768837082 Chr.17: 760130-760130 on Build GRCh38
rs77166377 Chr.17: 760232-760232 on Build GRCh38
rs763696517 Chr.17: 760183-760183 on Build GRCh38
rs74757321 Chr.17: 760227-760227 on Build GRCh38
rs1017481078 Chr.17: 760125-760125 on Build GRCh38
rs749600575 Chr.17: 760237-760237 on Build GRCh38
rs773581521 Chr.17: 760153-760153 on Build GRCh38
rs746710476 Chr.17: 760252-760252 on Build GRCh38
rs372139094 Chr.17: 760241-760241 on Build GRCh38
rs908481951 Chr.17: 760295-760295 on Build GRCh38
rs878875647 Chr.17: 760280-760280 on Build GRCh38
rs370921730 Chr.17: 760136-760136 on Build GRCh38
rs761867438 Chr.17: 760195-760195 on Build GRCh38
rs781648263 Chr.17: 760132-760132 on Build GRCh38
rs373779509 Chr.17: 760144-760144 on Build GRCh38
rs780127372 Chr.17: 760225-760225 on Build GRCh38
rs756156314 Chr.17: 760167-760167 on Build GRCh38
rs1009143797 Chr.17: 760158-760158 on Build GRCh38
rs1029167199 Chr.17: 760139-760139 on Build GRCh38
rs202190420 Chr.17: 760249-760249 on Build GRCh38

View Full Product Details