Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44170713-44170950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
UniGene
Hs.534517
Gene Chromosome Location
Chr.17: 44170706-44179083 on Build GRCh38
Transcripts
NM_080863.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4644070 c.89G>A p.R30Q Chr.17: 44170878-44170878 on Build GRCh38
4655257 c.83G>A p.R28Q Chr.17: 44170872-44170872 on Build GRCh38
706185 c.101A>T p.Q34L Chr.17: 44170890-44170890 on Build GRCh38
5508973 c.101A>C p.Q34P Chr.17: 44170890-44170890 on Build GRCh38
2149284 c.71_73delAAT p.E24_W25>G Chr.17: 44170860-44170862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776397394 Chr.17: 44170788-44170788 on Build GRCh38
rs771660999 Chr.17: 44170908-44170908 on Build GRCh38
rs56265171 Chr.17: 44170837-44170837 on Build GRCh38
rs953114278 Chr.17: 44170862-44170862 on Build GRCh38
rs144140141 Chr.17: 44170839-44170839 on Build GRCh38
rs763502506 Chr.17: 44170877-44170877 on Build GRCh38
rs751177767 Chr.17: 44170745-44170745 on Build GRCh38
rs200827398 Chr.17: 44170782-44170782 on Build GRCh38
rs140506716 Chr.17: 44170870-44170870 on Build GRCh38
rs761296640 Chr.17: 44170860-44170860 on Build GRCh38
rs945211411 Chr.17: 44170797-44170797 on Build GRCh38
rs144229400 Chr.17: 44170871-44170871 on Build GRCh38
rs766933192 Chr.17: 44170779-44170779 on Build GRCh38
rs367837547 Chr.17: 44170817-44170817 on Build GRCh38
rs759914571 Chr.17: 44170826-44170826 on Build GRCh38
rs745726300 Chr.17: 44170793-44170793 on Build GRCh38
rs192855432 Chr.17: 44170768-44170768 on Build GRCh38
rs777744675 Chr.17: 44170770-44170770 on Build GRCh38
rs759040207 Chr.17: 44170914-44170915 on Build GRCh38
rs777886270 Chr.17: 44170833-44170833 on Build GRCh38
rs201364601 Chr.17: 44170853-44170853 on Build GRCh38
rs769577332 Chr.17: 44170794-44170794 on Build GRCh38
rs375392717 Chr.17: 44170732-44170732 on Build GRCh38
rs767339837 Chr.17: 44170821-44170821 on Build GRCh38
rs749221208 Chr.17: 44170847-44170847 on Build GRCh38
rs752107616 Chr.17: 44170881-44170881 on Build GRCh38
rs768528706 Chr.17: 44170849-44170849 on Build GRCh38
rs775918008 Chr.17: 44170926-44170926 on Build GRCh38
rs754307863 Chr.17: 44170754-44170754 on Build GRCh38
rs778649100 Chr.17: 44170905-44170905 on Build GRCh38
rs149170804 Chr.17: 44170928-44170928 on Build GRCh38
rs767555969 Chr.17: 44170932-44170932 on Build GRCh38
rs754251112 Chr.17: 44170822-44170822 on Build GRCh38
rs528495680 Chr.17: 44170898-44170898 on Build GRCh38
rs773776656 Chr.17: 44170808-44170808 on Build GRCh38
rs760380688 Chr.17: 44170915-44170915 on Build GRCh38
rs768638576 Chr.17: 44170904-44170904 on Build GRCh38
rs748110246 Chr.17: 44170907-44170907 on Build GRCh38
rs369796709 Chr.17: 44170744-44170744 on Build GRCh38
rs773252427 Chr.17: 44170803-44170808 on Build GRCh38
rs146524979 Chr.17: 44170844-44170844 on Build GRCh38
rs376996083 Chr.17: 44170899-44170899 on Build GRCh38
rs781322387 Chr.17: 44170838-44170838 on Build GRCh38
rs765664033 Chr.17: 44170872-44170872 on Build GRCh38
rs199735813 Chr.17: 44170773-44170773 on Build GRCh38
rs758866778 Chr.17: 44170830-44170830 on Build GRCh38
rs143324294 Chr.17: 44170931-44170931 on Build GRCh38
rs747221966 Chr.17: 44170775-44170775 on Build GRCh38
rs145226168 Chr.17: 44170914-44170914 on Build GRCh38
rs568896513 Chr.17: 44170801-44170801 on Build GRCh38
rs766824314 Chr.17: 44170753-44170753 on Build GRCh38
rs755528606 Chr.17: 44170755-44170755 on Build GRCh38
rs762817455 Chr.17: 44170803-44170803 on Build GRCh38
rs764528773 Chr.17: 44170929-44170929 on Build GRCh38
rs763623202 Chr.17: 44170804-44170804 on Build GRCh38
rs779630328 Chr.17: 44170759-44170759 on Build GRCh38
rs770798712 Chr.17: 44170815-44170817 on Build GRCh38
rs761508752 Chr.17: 44170813-44170813 on Build GRCh38
rs779878771 Chr.17: 44170902-44170902 on Build GRCh38
rs200273394 Chr.17: 44170750-44170750 on Build GRCh38
rs749287866 Chr.17: 44170813-44170821 on Build GRCh38
rs781268578 Chr.17: 44170891-44170891 on Build GRCh38
rs377163601 Chr.17: 44170834-44170834 on Build GRCh38
rs534308231 Chr.17: 44170882-44170882 on Build GRCh38
rs533182208 Chr.17: 44170829-44170829 on Build GRCh38
rs370158958 Chr.17: 44170925-44170925 on Build GRCh38
rs764575988 Chr.17: 44170878-44170878 on Build GRCh38
rs774231569 Chr.17: 44170868-44170910 on Build GRCh38
rs753051378 Chr.17: 44170874-44170874 on Build GRCh38
rs905098190 Chr.17: 44170736-44170736 on Build GRCh38
rs767211105 Chr.17: 44170920-44170920 on Build GRCh38

View Full Product Details