Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27828742-27828993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSG1L
Entrez Gene ID
146395
Gene Name
GSG1 like
Gene Aliases
PRO19651
UniGene
Hs.91910
Gene Chromosome Location
Chr.16: 27787529-28063509 on Build GRCh38
Transcripts
NM_001109763.1 NM_001323900.1 NM_001323901.1 NM_144675.2 XM_011545750.2 XM_011545751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7333516 c.246G>A p.T82T Chr.16: 27828908-27828908 on Build GRCh38
4059708 c.808G>A p.A270T Chr.16: 27828811-27828811 on Build GRCh38
7370455 c.254C>G p.S85C Chr.16: 27828900-27828900 on Build GRCh38
6522719 c.683C>T p.T228I Chr.16: 27828936-27828936 on Build GRCh38
6143916 c.338C>A p.P113H Chr.16: 27828816-27828816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867536559 Chr.16: 27828838-27828838 on Build GRCh38
rs779216438 Chr.16: 27828796-27828796 on Build GRCh38
rs117918036 Chr.16: 27828950-27828950 on Build GRCh38
rs747518984 Chr.16: 27828820-27828820 on Build GRCh38
rs970706487 Chr.16: 27828866-27828866 on Build GRCh38
rs772785003 Chr.16: 27828840-27828840 on Build GRCh38
rs772235420 Chr.16: 27828806-27828806 on Build GRCh38
rs751413257 Chr.16: 27828769-27828769 on Build GRCh38
rs746938811 Chr.16: 27828779-27828779 on Build GRCh38
rs138692711 Chr.16: 27828966-27828966 on Build GRCh38
rs754827843 Chr.16: 27828776-27828776 on Build GRCh38
rs769451734 Chr.16: 27828832-27828832 on Build GRCh38
rs201582359 Chr.16: 27828823-27828823 on Build GRCh38
rs752444585 Chr.16: 27828874-27828874 on Build GRCh38
rs149224734 Chr.16: 27828831-27828831 on Build GRCh38
rs144726696 Chr.16: 27828908-27828908 on Build GRCh38
rs747105564 Chr.16: 27828909-27828909 on Build GRCh38
rs769099207 Chr.16: 27828911-27828911 on Build GRCh38
rs986799718 Chr.16: 27828932-27828932 on Build GRCh38
rs753737858 Chr.16: 27828830-27828830 on Build GRCh38
rs984428344 Chr.16: 27828867-27828867 on Build GRCh38
rs200764343 Chr.16: 27828860-27828860 on Build GRCh38
rs1024889153 Chr.16: 27828773-27828773 on Build GRCh38
rs750620355 Chr.16: 27828888-27828888 on Build GRCh38
rs772485041 Chr.16: 27828809-27828809 on Build GRCh38
rs142434117 Chr.16: 27828945-27828945 on Build GRCh38
rs751063139 Chr.16: 27828864-27828864 on Build GRCh38
rs748467525 Chr.16: 27828921-27828921 on Build GRCh38
rs759434857 Chr.16: 27828865-27828865 on Build GRCh38
rs780902974 Chr.16: 27828779-27828779 on Build GRCh38
rs553614382 Chr.16: 27828912-27828912 on Build GRCh38
rs371240922 Chr.16: 27828887-27828887 on Build GRCh38
rs376358621 Chr.16: 27828795-27828795 on Build GRCh38
rs551237467 Chr.16: 27828841-27828841 on Build GRCh38
rs758671274 Chr.16: 27828896-27828896 on Build GRCh38
rs745942755 Chr.16: 27828800-27828800 on Build GRCh38
rs748992322 Chr.16: 27828956-27828956 on Build GRCh38
rs550662906 Chr.16: 27828767-27828767 on Build GRCh38
rs767489098 Chr.16: 27828873-27828873 on Build GRCh38
rs752508560 Chr.16: 27828791-27828791 on Build GRCh38
rs780728755 Chr.16: 27828812-27828812 on Build GRCh38
rs755903809 Chr.16: 27828880-27828880 on Build GRCh38

View Full Product Details