Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35178892-35179161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREM
Entrez Gene ID
1390
Gene Name
cAMP responsive element modulator
Gene Aliases
CREM-2, ICER, hCREM-2
UniGene
Hs.200250
Gene Chromosome Location
Chr.10: 35126841-35212958 on Build GRCh38
Transcripts
NM_001267562.1 NM_001267563.1 NM_001267564.1 NM_001267565.1 NM_001267566.1 NM_001267567.1 NM_001881.3 NM_181571.2 NM_182769.2 NM_182770.2 NM_182771.1 NM_182772.1 NM_183011.1 NM_183012.1 NM_183013.2 NM_183060.2 NR_051973.1 NR_051974.1 NR_051975.1 XM_006717378.3 XM_006717379.3 XM_006717382.3 XM_006717383.3 XM_006717387.3 XM_011519316.2 XM_011519324.2 XM_011519325.2 XM_011519327.2 XM_011519330.2 XM_011519331.2 XM_011519332.2 XM_011519333.1 XM_011519335.2 XM_017015716.1 XM_017015717.1 XM_017015718.1 XM_017015719.1 XM_017015720.1 XM_017015721.1 XM_017015722.1 XM_017015723.1 XM_017015724.1 XM_017015725.1 XM_017015726.1 XM_017015727.1 XM_017015728.1 XM_017015729.1 XM_017015730.1 XM_017015731.1 XM_017015732.1 XM_017015733.1 XM_017015734.1 XM_017015735.1 XM_017015736.1 XM_017015737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7234267 c.267-1G>A p.? Chr.10: 35179133-35179133 on Build GRCh38
7147926 c.148G>C p.E50Q Chr.10: 35178958-35178958 on Build GRCh38
5616453 c.159A>G p.S53S Chr.10: 35178927-35178927 on Build GRCh38
4485073 c.198C>T p.S66S Chr.10: 35179155-35179155 on Build GRCh38
4485072 c.288C>T p.S96S Chr.10: 35179155-35179155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031206086 Chr.10: 35178958-35178958 on Build GRCh38
rs897614789 Chr.10: 35179064-35179064 on Build GRCh38
rs17854162 Chr.10: 35178929-35178929 on Build GRCh38
rs34556873 Chr.10: 35179061-35179062 on Build GRCh38
rs760555673 Chr.10: 35179107-35179107 on Build GRCh38
rs756392835 Chr.10: 35178969-35178969 on Build GRCh38
rs140809313 Chr.10: 35178915-35178915 on Build GRCh38
rs763817787 Chr.10: 35178975-35178975 on Build GRCh38
rs894621806 Chr.10: 35179098-35179098 on Build GRCh38
rs760608530 Chr.10: 35178971-35178971 on Build GRCh38
rs771541954 Chr.10: 35179086-35179086 on Build GRCh38
rs774875136 Chr.10: 35179092-35179092 on Build GRCh38
rs1035984529 Chr.10: 35179031-35179031 on Build GRCh38
rs369751854 Chr.10: 35179109-35179109 on Build GRCh38
rs770328240 Chr.10: 35179035-35179035 on Build GRCh38
rs766990868 Chr.10: 35178968-35178968 on Build GRCh38
rs747464101 Chr.10: 35179004-35179004 on Build GRCh38
rs766749175 Chr.10: 35179041-35179041 on Build GRCh38
rs761281142 Chr.10: 35179017-35179018 on Build GRCh38
rs372250022 Chr.10: 35179034-35179034 on Build GRCh38
rs892125898 Chr.10: 35178961-35178961 on Build GRCh38
rs368315723 Chr.10: 35178984-35178984 on Build GRCh38
rs527347965 Chr.10: 35179014-35179014 on Build GRCh38
rs769077047 Chr.10: 35178919-35178919 on Build GRCh38
rs938411422 Chr.10: 35179066-35179066 on Build GRCh38
rs757430447 Chr.10: 35178983-35178983 on Build GRCh38
rs774036879 Chr.10: 35179083-35179083 on Build GRCh38
rs1057484763 Chr.10: 35179072-35179072 on Build GRCh38
rs780005539 Chr.10: 35179002-35179002 on Build GRCh38
rs188189426 Chr.10: 35178985-35178985 on Build GRCh38
rs777152673 Chr.10: 35178937-35178937 on Build GRCh38
rs759174978 Chr.10: 35179084-35179084 on Build GRCh38
rs753655454 Chr.10: 35178978-35178978 on Build GRCh38
rs748496572 Chr.10: 35179026-35179026 on Build GRCh38
rs758501977 Chr.10: 35178990-35178990 on Build GRCh38
rs777182145 Chr.10: 35179082-35179083 on Build GRCh38
rs567237282 Chr.10: 35179047-35179047 on Build GRCh38
rs201032782 Chr.10: 35178952-35178952 on Build GRCh38
rs762647846 Chr.10: 35178941-35178941 on Build GRCh38
rs759061714 Chr.10: 35178953-35178953 on Build GRCh38
rs764048891 Chr.10: 35179113-35179113 on Build GRCh38
rs547427073 Chr.10: 35179018-35179018 on Build GRCh38
rs150152061 Chr.10: 35178927-35178927 on Build GRCh38
rs770743308 Chr.10: 35178950-35178950 on Build GRCh38

View Full Product Details