Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47372824-47373081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK1
Entrez Gene ID
51727
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
UniGene
Hs.731647
Gene Chromosome Location
Chr.1: 47333797-47378839 on Build GRCh38
Transcripts
NM_001136140.1 NM_016308.2 NR_046394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2191781 c.326A>T p.D109V Chr.1: 47372962-47372962 on Build GRCh38
910429 c.363C>A p.F121L Chr.1: 47372999-47372999 on Build GRCh38
2191783 c.354G>T p.K118N Chr.1: 47372990-47372990 on Build GRCh38
146505 c.344A>G p.N115S Chr.1: 47372980-47372980 on Build GRCh38
4428419 c.331A>G p.T111A Chr.1: 47372967-47372967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022000350 Chr.1: 47372946-47372946 on Build GRCh38
rs572636928 Chr.1: 47372937-47372937 on Build GRCh38
rs376110727 Chr.1: 47372979-47372979 on Build GRCh38
rs762579970 Chr.1: 47373049-47373049 on Build GRCh38
rs763787616 Chr.1: 47373052-47373052 on Build GRCh38
rs768826233 Chr.1: 47372932-47372932 on Build GRCh38
rs749376825 Chr.1: 47372924-47372924 on Build GRCh38
rs748442752 Chr.1: 47372949-47372949 on Build GRCh38
rs977859310 Chr.1: 47372973-47372973 on Build GRCh38
rs72553946 Chr.1: 47372955-47372955 on Build GRCh38
rs756142995 Chr.1: 47372905-47372905 on Build GRCh38
rs1054097379 Chr.1: 47373022-47373022 on Build GRCh38
rs1017464709 Chr.1: 47372882-47372882 on Build GRCh38
rs113396807 Chr.1: 47372874-47372875 on Build GRCh38
rs373156341 Chr.1: 47372868-47372868 on Build GRCh38
rs775071933 Chr.1: 47372986-47372986 on Build GRCh38
rs758781720 Chr.1: 47372920-47372922 on Build GRCh38
rs72553947 Chr.1: 47372980-47372980 on Build GRCh38
rs201129706 Chr.1: 47372919-47372919 on Build GRCh38
rs900192982 Chr.1: 47372915-47372915 on Build GRCh38
rs780053127 Chr.1: 47372910-47372910 on Build GRCh38
rs892812530 Chr.1: 47373041-47373041 on Build GRCh38
rs375212744 Chr.1: 47372952-47372952 on Build GRCh38
rs960886197 Chr.1: 47372994-47372994 on Build GRCh38
rs751273816 Chr.1: 47373057-47373057 on Build GRCh38

View Full Product Details