Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5234257-5234488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBD
Entrez Gene ID
3045
Gene Name
hemoglobin subunit delta
Gene Aliases
-
UniGene
Hs.699280
Gene Chromosome Location
Chr.11: 5232829-5234628 on Build GRCh38
Transcripts
NM_000519.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
689096 c.54A>C p.K18N Chr.11: 5234380-5234380 on Build GRCh38
4455834 c.91A>C p.R31R Chr.11: 5234343-5234343 on Build GRCh38
4689551 c.36C>A p.V12V Chr.11: 5234398-5234398 on Build GRCh38
4033516 c.92G>A p.R31K Chr.11: 5234342-5234342 on Build GRCh38
3375835 c.90C>A p.G30G Chr.11: 5234344-5234344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35433207 Chr.11: 5234426-5234426 on Build GRCh38
rs757622703 Chr.11: 5234387-5234387 on Build GRCh38
rs376989241 Chr.11: 5234349-5234349 on Build GRCh38
rs1010664704 Chr.11: 5234286-5234286 on Build GRCh38
rs886048395 Chr.11: 5234358-5234358 on Build GRCh38
rs779306538 Chr.11: 5234432-5234432 on Build GRCh38
rs777991732 Chr.11: 5234374-5234374 on Build GRCh38
rs780459430 Chr.11: 5234450-5234450 on Build GRCh38
rs562543884 Chr.11: 5234418-5234418 on Build GRCh38
rs780686206 Chr.11: 5234361-5234361 on Build GRCh38
rs35152987 Chr.11: 5234352-5234352 on Build GRCh38
rs746381682 Chr.11: 5234392-5234392 on Build GRCh38
rs779230012 Chr.11: 5234391-5234391 on Build GRCh38
rs759483345 Chr.11: 5234417-5234417 on Build GRCh38
rs991672387 Chr.11: 5234447-5234447 on Build GRCh38
rs762782488 Chr.11: 5234405-5234405 on Build GRCh38
rs768225600 Chr.11: 5234314-5234314 on Build GRCh38
rs34991152 Chr.11: 5234429-5234429 on Build GRCh38
rs34090605 Chr.11: 5234399-5234399 on Build GRCh38
rs762875104 Chr.11: 5234338-5234338 on Build GRCh38
rs769446735 Chr.11: 5234459-5234459 on Build GRCh38
rs749699838 Chr.11: 5234292-5234292 on Build GRCh38
rs281864510 Chr.11: 5234427-5234427 on Build GRCh38
rs747684647 Chr.11: 5234396-5234396 on Build GRCh38
rs281864493 Chr.11: 5234431-5234431 on Build GRCh38
rs1018809713 Chr.11: 5234358-5234358 on Build GRCh38
rs35291639 Chr.11: 5234393-5234393 on Build GRCh38
rs751533248 Chr.11: 5234351-5234351 on Build GRCh38
rs564626562 Chr.11: 5234436-5234436 on Build GRCh38
rs369305779 Chr.11: 5234373-5234373 on Build GRCh38
rs34093840 Chr.11: 5234372-5234372 on Build GRCh38
rs35654785 Chr.11: 5234342-5234342 on Build GRCh38
rs773007023 Chr.11: 5234401-5234401 on Build GRCh38
rs35395083 Chr.11: 5234366-5234366 on Build GRCh38
rs1054451119 Chr.11: 5234285-5234285 on Build GRCh38
rs776071703 Chr.11: 5234317-5234317 on Build GRCh38
rs761527836 Chr.11: 5234320-5234320 on Build GRCh38
rs281864565 Chr.11: 5234415-5234415 on Build GRCh38
rs34389944 Chr.11: 5234357-5234357 on Build GRCh38
rs750803605 Chr.11: 5234435-5234435 on Build GRCh38
rs35345203 Chr.11: 5234409-5234409 on Build GRCh38
rs752791038 Chr.11: 5234323-5234324 on Build GRCh38
rs34313675 Chr.11: 5234395-5234395 on Build GRCh38
rs764233236 Chr.11: 5234422-5234422 on Build GRCh38
rs35540211 Chr.11: 5234340-5234340 on Build GRCh38
rs774077053 Chr.11: 5234413-5234413 on Build GRCh38
rs1029882418 Chr.11: 5234310-5234310 on Build GRCh38
rs774952470 Chr.11: 5234302-5234302 on Build GRCh38
rs773095083 Chr.11: 5234329-5234329 on Build GRCh38
rs63751203 Chr.11: 5234411-5234411 on Build GRCh38
rs747597296 Chr.11: 5234456-5234456 on Build GRCh38
rs34012192 Chr.11: 5234385-5234385 on Build GRCh38
rs770750995 Chr.11: 5234406-5234406 on Build GRCh38
rs772497026 Chr.11: 5234394-5234394 on Build GRCh38
rs372352398 Chr.11: 5234325-5234325 on Build GRCh38
rs746437527 Chr.11: 5234303-5234303 on Build GRCh38
rs533796952 Chr.11: 5234301-5234301 on Build GRCh38
rs34460332 Chr.11: 5234360-5234360 on Build GRCh38
rs758204272 Chr.11: 5234433-5234439 on Build GRCh38
rs34289459 Chr.11: 5234353-5234353 on Build GRCh38
rs281864508 Chr.11: 5234410-5234410 on Build GRCh38
rs35406175 Chr.11: 5234420-5234420 on Build GRCh38

View Full Product Details