Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20368942-20369187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSM1
Entrez Gene ID
3642
Gene Name
INSM transcriptional repressor 1
Gene Aliases
IA-1, IA1
UniGene
Hs.89584
Gene Chromosome Location
Chr.20: 20368121-20370949 on Build GRCh38
Transcripts
NM_002196.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029558 c.920C>G p.P307R Chr.20: 20369187-20369187 on Build GRCh38
7372298 c.802A>G p.I268V Chr.20: 20369069-20369069 on Build GRCh38
321023 c.853C>A p.H285N Chr.20: 20369120-20369120 on Build GRCh38
4692655 c.775_779delGGCGC p.R262fs*75 Chr.20: 20369042-20369046 on Build GRCh38
6414140 c.861C>A p.C287* Chr.20: 20369128-20369128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778760230 Chr.20: 20369025-20369025 on Build GRCh38
rs761942287 Chr.20: 20369047-20369047 on Build GRCh38
rs753044882 Chr.20: 20369085-20369085 on Build GRCh38
rs755453728 Chr.20: 20369080-20369080 on Build GRCh38
rs775273451 Chr.20: 20369128-20369128 on Build GRCh38
rs150281189 Chr.20: 20369169-20369169 on Build GRCh38
rs769561074 Chr.20: 20369045-20369045 on Build GRCh38
rs767489803 Chr.20: 20368970-20368970 on Build GRCh38
rs756762929 Chr.20: 20369110-20369110 on Build GRCh38
rs769648955 Chr.20: 20369122-20369122 on Build GRCh38
rs866257491 Chr.20: 20368985-20368985 on Build GRCh38
rs750490137 Chr.20: 20369057-20369057 on Build GRCh38
rs535185500 Chr.20: 20369113-20369113 on Build GRCh38
rs767716364 Chr.20: 20369049-20369049 on Build GRCh38
rs776513961 Chr.20: 20369043-20369043 on Build GRCh38
rs770799634 Chr.20: 20369040-20369040 on Build GRCh38
rs749063014 Chr.20: 20369131-20369131 on Build GRCh38
rs747790767 Chr.20: 20369027-20369027 on Build GRCh38
rs766833776 Chr.20: 20369069-20369069 on Build GRCh38
rs375892105 Chr.20: 20368988-20368988 on Build GRCh38
rs780558061 Chr.20: 20368990-20368990 on Build GRCh38
rs759369820 Chr.20: 20369044-20369044 on Build GRCh38
rs781162050 Chr.20: 20369116-20369116 on Build GRCh38
rs746535687 Chr.20: 20369102-20369102 on Build GRCh38
rs567774754 Chr.20: 20369021-20369021 on Build GRCh38
rs746473643 Chr.20: 20369036-20369036 on Build GRCh38
rs760767599 Chr.20: 20369065-20369065 on Build GRCh38
rs771680899 Chr.20: 20369028-20369028 on Build GRCh38
rs772214127 Chr.20: 20369132-20369132 on Build GRCh38
rs755361688 Chr.20: 20369022-20369022 on Build GRCh38
rs745797958 Chr.20: 20369119-20369119 on Build GRCh38
rs886069759 Chr.20: 20369138-20369138 on Build GRCh38
rs549579560 Chr.20: 20369014-20369014 on Build GRCh38
rs773452034 Chr.20: 20369133-20369133 on Build GRCh38
rs867257710 Chr.20: 20369012-20369012 on Build GRCh38
rs758004234 Chr.20: 20369086-20369086 on Build GRCh38
rs750450862 Chr.20: 20368984-20368984 on Build GRCh38
rs867487486 Chr.20: 20368961-20368961 on Build GRCh38
rs774619030 Chr.20: 20369046-20369046 on Build GRCh38
rs777437175 Chr.20: 20369101-20369101 on Build GRCh38

View Full Product Details