Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50549219-50549444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHDC7B
Entrez Gene ID
113730
Gene Name
kelch domain containing 7B
Gene Aliases
-
UniGene
Hs.137007
Gene Chromosome Location
Chr.22: 50548033-50551023 on Build GRCh38
Transcripts
NM_138433.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004882 c.1108G>A p.V370I Chr.22: 50549274-50549274 on Build GRCh38
6024507 c.780_781insG p.I263fs*226 Chr.22: 50549243-50549244 on Build GRCh38
6696382 c.1077delG p.G361fs*21 Chr.22: 50549243-50549243 on Build GRCh38
6346765 c.1186G>T p.A396S Chr.22: 50549352-50549352 on Build GRCh38
4105222 c.1263C>T p.Y421Y Chr.22: 50549429-50549429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376479128 Chr.22: 50549323-50549323 on Build GRCh38
rs1024760528 Chr.22: 50549378-50549378 on Build GRCh38
rs373269356 Chr.22: 50549312-50549312 on Build GRCh38
rs956147662 Chr.22: 50549393-50549393 on Build GRCh38
rs745950415 Chr.22: 50549286-50549286 on Build GRCh38
rs373321869 Chr.22: 50549257-50549257 on Build GRCh38
rs149296076 Chr.22: 50549373-50549373 on Build GRCh38
rs994660574 Chr.22: 50549315-50549315 on Build GRCh38
rs1039251947 Chr.22: 50549411-50549411 on Build GRCh38
rs779277541 Chr.22: 50549382-50549382 on Build GRCh38
rs943236885 Chr.22: 50549403-50549403 on Build GRCh38
rs755486026 Chr.22: 50549243-50549243 on Build GRCh38
rs768538997 Chr.22: 50549300-50549300 on Build GRCh38
rs369384415 Chr.22: 50549304-50549304 on Build GRCh38
rs751049025 Chr.22: 50549242-50549242 on Build GRCh38
rs773997042 Chr.22: 50549425-50549425 on Build GRCh38
rs756445154 Chr.22: 50549250-50549250 on Build GRCh38
rs769527090 Chr.22: 50549314-50549314 on Build GRCh38
rs139536561 Chr.22: 50549318-50549318 on Build GRCh38
rs61746062 Chr.22: 50549414-50549414 on Build GRCh38
rs757615328 Chr.22: 50549375-50549375 on Build GRCh38
rs776456614 Chr.22: 50549290-50549292 on Build GRCh38
rs1050477074 Chr.22: 50549253-50549253 on Build GRCh38
rs748467126 Chr.22: 50549248-50549248 on Build GRCh38
rs147168342 Chr.22: 50549261-50549261 on Build GRCh38
rs765993261 Chr.22: 50549332-50549332 on Build GRCh38
rs753201278 Chr.22: 50549350-50549350 on Build GRCh38
rs751013797 Chr.22: 50549259-50549259 on Build GRCh38
rs769685749 Chr.22: 50549402-50549402 on Build GRCh38
rs751088145 Chr.22: 50549333-50549333 on Build GRCh38
rs759020961 Chr.22: 50549348-50549348 on Build GRCh38
rs767923639 Chr.22: 50549349-50549349 on Build GRCh38
rs957546181 Chr.22: 50549417-50549417 on Build GRCh38
rs375448641 Chr.22: 50549274-50549274 on Build GRCh38
rs370876870 Chr.22: 50549387-50549387 on Build GRCh38
rs781720677 Chr.22: 50549246-50549246 on Build GRCh38
rs756579554 Chr.22: 50549357-50549357 on Build GRCh38
rs924588273 Chr.22: 50549374-50549374 on Build GRCh38
rs748075949 Chr.22: 50549397-50549397 on Build GRCh38
rs574329986 Chr.22: 50549408-50549408 on Build GRCh38
rs778325249 Chr.22: 50549368-50549368 on Build GRCh38
rs117231091 Chr.22: 50549396-50549396 on Build GRCh38

View Full Product Details