Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144078604-144078865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC4
Entrez Gene ID
54512
Gene Name
exosome component 4
Gene Aliases
RRP41, RRP41A, Rrp41p, SKI6, Ski6p, hRrp41p, p12A
UniGene
Hs.632041
Gene Chromosome Location
Chr.8: 144064092-144080648 on Build GRCh38
Transcripts
NM_019037.2 XM_011517134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205843 c.88G>T p.G30C Chr.8: 144078816-144078816 on Build GRCh38
6837378 c.13G>C p.E5Q Chr.8: 144078741-144078741 on Build GRCh38
5553432 c.124G>A p.E42K Chr.8: 144078852-144078852 on Build GRCh38
1489119 c.7G>A p.G3R Chr.8: 144078735-144078735 on Build GRCh38
7152358 c.121A>G p.I41V Chr.8: 144078849-144078849 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782160353 Chr.8: 144078743-144078743 on Build GRCh38
rs782658839 Chr.8: 144078696-144078696 on Build GRCh38
rs941096219 Chr.8: 144078721-144078721 on Build GRCh38
rs779541092 Chr.8: 144078758-144078758 on Build GRCh38
rs781965076 Chr.8: 144078819-144078819 on Build GRCh38
rs782335169 Chr.8: 144078733-144078733 on Build GRCh38
rs371352092 Chr.8: 144078839-144078839 on Build GRCh38
rs781890149 Chr.8: 144078783-144078783 on Build GRCh38
rs782647207 Chr.8: 144078785-144078785 on Build GRCh38
rs367830770 Chr.8: 144078788-144078788 on Build GRCh38
rs541379674 Chr.8: 144078697-144078697 on Build GRCh38
rs559257152 Chr.8: 144078667-144078667 on Build GRCh38
rs558461285 Chr.8: 144078632-144078632 on Build GRCh38
rs781995308 Chr.8: 144078752-144078752 on Build GRCh38
rs782660594 Chr.8: 144078803-144078803 on Build GRCh38
rs143107280 Chr.8: 144078842-144078842 on Build GRCh38
rs34798795 Chr.8: 144078770-144078770 on Build GRCh38
rs782427234 Chr.8: 144078801-144078801 on Build GRCh38
rs76352871 Chr.8: 144078625-144078625 on Build GRCh38
rs915223927 Chr.8: 144078651-144078651 on Build GRCh38
rs781904161 Chr.8: 144078771-144078771 on Build GRCh38
rs781929052 Chr.8: 144078741-144078741 on Build GRCh38
rs782487570 Chr.8: 144078787-144078787 on Build GRCh38
rs576410912 Chr.8: 144078624-144078624 on Build GRCh38
rs782596773 Chr.8: 144078686-144078686 on Build GRCh38
rs782255719 Chr.8: 144078812-144078812 on Build GRCh38
rs781840523 Chr.8: 144078786-144078786 on Build GRCh38
rs572024698 Chr.8: 144078695-144078695 on Build GRCh38
rs781807786 Chr.8: 144078689-144078689 on Build GRCh38
rs371109781 Chr.8: 144078708-144078708 on Build GRCh38
rs781849408 Chr.8: 144078678-144078678 on Build GRCh38
rs540903907 Chr.8: 144078648-144078648 on Build GRCh38
rs574414438 Chr.8: 144078681-144078681 on Build GRCh38
rs782077905 Chr.8: 144078828-144078828 on Build GRCh38
rs782631281 Chr.8: 144078713-144078713 on Build GRCh38
rs781819833 Chr.8: 144078765-144078765 on Build GRCh38
rs782514177 Chr.8: 144078784-144078784 on Build GRCh38
rs782699055 Chr.8: 144078780-144078780 on Build GRCh38
rs782132080 Chr.8: 144078698-144078698 on Build GRCh38
rs978306897 Chr.8: 144078664-144078664 on Build GRCh38
rs781833704 Chr.8: 144078659-144078659 on Build GRCh38
rs577014328 Chr.8: 144078634-144078634 on Build GRCh38
rs934258599 Chr.8: 144078685-144078685 on Build GRCh38
rs782403640 Chr.8: 144078751-144078751 on Build GRCh38
rs782108842 Chr.8: 144078755-144078755 on Build GRCh38
rs782365848 Chr.8: 144078813-144078813 on Build GRCh38
rs782218536 Chr.8: 144078717-144078717 on Build GRCh38
rs781919685 Chr.8: 144078841-144078841 on Build GRCh38

View Full Product Details