Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75961117-75961344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDAD1
Entrez Gene ID
55153
Gene Name
SDA1 domain containing 1
Gene Aliases
-
UniGene
Hs.632604
Gene Chromosome Location
Chr.4: 75949906-75990962 on Build GRCh38
Transcripts
NM_001288983.1 NM_001288984.1 NM_018115.3 XM_005263102.4 XM_005263105.4 XM_017008354.1 XM_017008355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431077 c.1237C>A p.L413I Chr.4: 75961253-75961253 on Build GRCh38
4125988 c.1190C>T p.A397V Chr.4: 75961300-75961300 on Build GRCh38
1310259 c.1187A>G p.N396S Chr.4: 75961303-75961303 on Build GRCh38
7095212 c.1242C>G p.L414L Chr.4: 75961248-75961248 on Build GRCh38
7111149 c.1185C>G p.I395M Chr.4: 75961305-75961305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775227985 Chr.4: 75961190-75961191 on Build GRCh38
rs369140120 Chr.4: 75961303-75961303 on Build GRCh38
rs772691754 Chr.4: 75961248-75961248 on Build GRCh38
rs1013554798 Chr.4: 75961149-75961149 on Build GRCh38
rs202155622 Chr.4: 75961285-75961285 on Build GRCh38
rs764734200 Chr.4: 75961276-75961276 on Build GRCh38
rs902036806 Chr.4: 75961151-75961151 on Build GRCh38
rs994060624 Chr.4: 75961166-75961166 on Build GRCh38
rs776293957 Chr.4: 75961265-75961265 on Build GRCh38
rs780221956 Chr.4: 75961191-75961191 on Build GRCh38
rs369181093 Chr.4: 75961256-75961256 on Build GRCh38
rs748623734 Chr.4: 75961244-75961244 on Build GRCh38
rs755018096 Chr.4: 75961302-75961302 on Build GRCh38
rs762938714 Chr.4: 75961280-75961280 on Build GRCh38
rs751344802 Chr.4: 75961180-75961180 on Build GRCh38
rs749381831 Chr.4: 75961206-75961206 on Build GRCh38
rs372045634 Chr.4: 75961259-75961259 on Build GRCh38
rs147216033 Chr.4: 75961222-75961222 on Build GRCh38
rs959356889 Chr.4: 75961299-75961299 on Build GRCh38
rs757028091 Chr.4: 75961290-75961290 on Build GRCh38
rs756398904 Chr.4: 75961183-75961183 on Build GRCh38
rs770755760 Chr.4: 75961263-75961263 on Build GRCh38
rs375343526 Chr.4: 75961271-75961271 on Build GRCh38
rs143777133 Chr.4: 75961221-75961221 on Build GRCh38
rs758191084 Chr.4: 75961174-75961174 on Build GRCh38
rs552023073 Chr.4: 75961284-75961284 on Build GRCh38
rs754051760 Chr.4: 75961298-75961298 on Build GRCh38
rs1025439377 Chr.4: 75961160-75961160 on Build GRCh38
rs377507179 Chr.4: 75961203-75961203 on Build GRCh38
rs752636974 Chr.4: 75961173-75961173 on Build GRCh38
rs113244163 Chr.4: 75961272-75961272 on Build GRCh38
rs766532305 Chr.4: 75961296-75961296 on Build GRCh38
rs777484376 Chr.4: 75961176-75961176 on Build GRCh38
rs775300867 Chr.4: 75961279-75961279 on Build GRCh38

View Full Product Details