Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182812057-182812316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPL
Entrez Gene ID
80896
Gene Name
N-acetylneuraminate pyruvate lyase
Gene Aliases
C112, C1orf13, NAL, NPL1
UniGene
Hs.496969
Gene Chromosome Location
Chr.1: 182789449-182830384 on Build GRCh38
Transcripts
NM_001200050.1 NM_001200051.1 NM_001200052.1 NM_001200056.1 NM_030769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4538596 c.256G>A p.G86R Chr.1: 182812181-182812181 on Build GRCh38
6323469 c.288G>A p.L96L Chr.1: 182812213-182812213 on Build GRCh38
4538594 c.256G>A p.G86R Chr.1: 182812181-182812181 on Build GRCh38
677706 c.253G>C p.V85L Chr.1: 182812178-182812178 on Build GRCh38
4538595 c.199G>A p.G67R Chr.1: 182812181-182812181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141892236 Chr.1: 182812178-182812178 on Build GRCh38
rs758017325 Chr.1: 182812214-182812214 on Build GRCh38
rs764820846 Chr.1: 182812210-182812210 on Build GRCh38
rs966156659 Chr.1: 182812093-182812093 on Build GRCh38
rs775114427 Chr.1: 182812244-182812244 on Build GRCh38
rs776351187 Chr.1: 182812134-182812134 on Build GRCh38
rs763152684 Chr.1: 182812159-182812159 on Build GRCh38
rs371975075 Chr.1: 182812128-182812128 on Build GRCh38
rs752009742 Chr.1: 182812163-182812163 on Build GRCh38
rs565831079 Chr.1: 182812225-182812225 on Build GRCh38
rs369943953 Chr.1: 182812123-182812123 on Build GRCh38
rs781696984 Chr.1: 182812231-182812231 on Build GRCh38
rs1027641747 Chr.1: 182812290-182812290 on Build GRCh38
rs777315527 Chr.1: 182812217-182812217 on Build GRCh38
rs764307290 Chr.1: 182812160-182812160 on Build GRCh38
rs1044481655 Chr.1: 182812211-182812211 on Build GRCh38
rs775729521 Chr.1: 182812153-182812153 on Build GRCh38
rs750756881 Chr.1: 182812198-182812198 on Build GRCh38
rs2225789 Chr.1: 182812138-182812138 on Build GRCh38
rs745669422 Chr.1: 182812237-182812237 on Build GRCh38
rs530819200 Chr.1: 182812288-182812288 on Build GRCh38
rs752355220 Chr.1: 182812212-182812212 on Build GRCh38
rs759375168 Chr.1: 182812136-182812136 on Build GRCh38
rs745941974 Chr.1: 182812242-182812242 on Build GRCh38
rs77446110 Chr.1: 182812124-182812124 on Build GRCh38
rs764401213 Chr.1: 182812121-182812122 on Build GRCh38
rs979126960 Chr.1: 182812129-182812129 on Build GRCh38
rs950147387 Chr.1: 182812193-182812193 on Build GRCh38
rs771522837 Chr.1: 182812111-182812111 on Build GRCh38
rs756443180 Chr.1: 182812209-182812209 on Build GRCh38
rs751438154 Chr.1: 182812177-182812177 on Build GRCh38
rs747625392 Chr.1: 182812105-182812105 on Build GRCh38
rs539763982 Chr.1: 182812235-182812235 on Build GRCh38
rs749568670 Chr.1: 182812261-182812261 on Build GRCh38
rs769588948 Chr.1: 182812240-182812240 on Build GRCh38
rs768063985 Chr.1: 182812187-182812187 on Build GRCh38
rs896335386 Chr.1: 182812191-182812191 on Build GRCh38
rs551653057 Chr.1: 182812270-182812270 on Build GRCh38

View Full Product Details