Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 62589563-62589794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAIN3
Entrez Gene ID
286183
Gene Name
Na+/K+ transporting ATPase interacting 3
Gene Aliases
FAM77D
UniGene
Hs.654662
Gene Chromosome Location
Chr.8: 62248942-62999652 on Build GRCh38
Transcripts
NM_001304533.1 NR_130764.1 XM_011517511.2 XM_011517512.2 XM_017013359.1 XM_017013360.1 XM_017013361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3951763 c.229G>T p.V77L Chr.8: 62589750-62589750 on Build GRCh38
7302329 c.257T>C p.V86A Chr.8: 62589778-62589778 on Build GRCh38
4185704 c.266T>C p.L89P Chr.8: 62589787-62589787 on Build GRCh38
6726509 c.223T>A p.W75R Chr.8: 62589744-62589744 on Build GRCh38
6113604 c.201G>T p.V67V Chr.8: 62589722-62589722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570989024 Chr.8: 62589708-62589708 on Build GRCh38
rs764108057 Chr.8: 62589693-62589693 on Build GRCh38
rs989977732 Chr.8: 62589669-62589669 on Build GRCh38
rs1045456179 Chr.8: 62589734-62589734 on Build GRCh38
rs945553422 Chr.8: 62589692-62589692 on Build GRCh38
rs552812324 Chr.8: 62589641-62589641 on Build GRCh38
rs377166648 Chr.8: 62589740-62589740 on Build GRCh38
rs958025166 Chr.8: 62589608-62589608 on Build GRCh38
rs537368359 Chr.8: 62589598-62589598 on Build GRCh38
rs943616365 Chr.8: 62589726-62589726 on Build GRCh38
rs980800830 Chr.8: 62589593-62589593 on Build GRCh38
rs758133009 Chr.8: 62589743-62589743 on Build GRCh38
rs750949067 Chr.8: 62589730-62589730 on Build GRCh38
rs754692326 Chr.8: 62589752-62589752 on Build GRCh38
rs758766040 Chr.8: 62589738-62589738 on Build GRCh38
rs762238314 Chr.8: 62589707-62589707 on Build GRCh38
rs373253501 Chr.8: 62589675-62589675 on Build GRCh38
rs776701464 Chr.8: 62589697-62589697 on Build GRCh38
rs370357908 Chr.8: 62589750-62589750 on Build GRCh38
rs757432875 Chr.8: 62589725-62589725 on Build GRCh38
rs2306401 Chr.8: 62589592-62589592 on Build GRCh38
rs775754266 Chr.8: 62589677-62589677 on Build GRCh38
rs770683056 Chr.8: 62589676-62589676 on Build GRCh38
rs913958551 Chr.8: 62589688-62589688 on Build GRCh38

View Full Product Details