Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 63391855-63392124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R5E
Entrez Gene ID
5529
Gene Name
protein phosphatase 2 regulatory subunit B'epsilon
Gene Aliases
-
UniGene
Hs.334868
Gene Chromosome Location
Chr.14: 63373892-63543361 on Build GRCh38
Transcripts
NM_001282179.1 NM_001282180.1 NM_001282181.1 NM_001282182.1 NM_006246.3 NR_104104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386619 c.862A>G p.I288V Chr.14: 63392013-63392013 on Build GRCh38
1707434 c.887C>T p.P296L Chr.14: 63391988-63391988 on Build GRCh38
6521564 c.912G>T p.R304S Chr.14: 63391859-63391859 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764344310 Chr.14: 63392017-63392017 on Build GRCh38
rs765723703 Chr.14: 63391917-63391917 on Build GRCh38
rs867114033 Chr.14: 63391932-63391932 on Build GRCh38
rs369213359 Chr.14: 63391916-63391916 on Build GRCh38
rs772679993 Chr.14: 63391989-63391989 on Build GRCh38
rs140406327 Chr.14: 63392020-63392020 on Build GRCh38
rs769176745 Chr.14: 63391944-63391944 on Build GRCh38
rs529594662 Chr.14: 63391950-63391950 on Build GRCh38
rs201121783 Chr.14: 63391937-63391937 on Build GRCh38
rs914430533 Chr.14: 63391883-63391883 on Build GRCh38
rs1012484270 Chr.14: 63391973-63391973 on Build GRCh38
rs761151881 Chr.14: 63392013-63392013 on Build GRCh38
rs371773001 Chr.14: 63392071-63392071 on Build GRCh38
rs765201624 Chr.14: 63392048-63392048 on Build GRCh38
rs192959225 Chr.14: 63391961-63391961 on Build GRCh38
rs1056694344 Chr.14: 63391964-63391964 on Build GRCh38
rs768261348 Chr.14: 63391887-63391887 on Build GRCh38
rs896742993 Chr.14: 63391965-63391965 on Build GRCh38
rs756812498 Chr.14: 63392034-63392038 on Build GRCh38
rs940421002 Chr.14: 63392086-63392086 on Build GRCh38
rs887988196 Chr.14: 63392077-63392077 on Build GRCh38
rs368121677 Chr.14: 63391943-63391943 on Build GRCh38
rs780830701 Chr.14: 63391918-63391918 on Build GRCh38
rs750566730 Chr.14: 63392052-63392052 on Build GRCh38
rs368179547 Chr.14: 63392010-63392010 on Build GRCh38

View Full Product Details