Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 132436582-132436855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPCML
Entrez Gene ID
4978
Gene Name
opioid binding protein/cell adhesion molecule like
Gene Aliases
IGLON1, OBCAM, OPCM
UniGene
Hs.4817
Gene Chromosome Location
Chr.11: 132403361-133532983 on Build GRCh38
Transcripts
NM_001012393.2 NM_001319103.1 NM_001319104.1 NM_001319105.1 NM_001319106.1 NM_002545.4 XM_006718846.2 XM_011542856.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1724168 c.665-11_665-10delGT p.? Chr.11: 132436789-132436790 on Build GRCh38
1578667 c.653A>C p.Y218S Chr.11: 132436770-132436770 on Build GRCh38
6068216 c.695C>T p.T232I Chr.11: 132436749-132436749 on Build GRCh38
1627872 c.744G>C p.Q248H Chr.11: 132436679-132436679 on Build GRCh38
4190005 c.670A>G p.N224D Chr.11: 132436753-132436753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746296988 Chr.11: 132436749-132436749 on Build GRCh38
rs117621063 Chr.11: 132436614-132436614 on Build GRCh38
rs752741994 Chr.11: 132436707-132436707 on Build GRCh38
rs773908755 Chr.11: 132436824-132436825 on Build GRCh38
rs367841063 Chr.11: 132436740-132436740 on Build GRCh38
rs776839773 Chr.11: 132436804-132436805 on Build GRCh38
rs192871884 Chr.11: 132436823-132436823 on Build GRCh38
rs779615860 Chr.11: 132436827-132436827 on Build GRCh38
rs145121891 Chr.11: 132436735-132436735 on Build GRCh38
rs766191726 Chr.11: 132436808-132436808 on Build GRCh38
rs754016747 Chr.11: 132436820-132436820 on Build GRCh38
rs755175009 Chr.11: 132436692-132436692 on Build GRCh38
rs142073080 Chr.11: 132436772-132436772 on Build GRCh38
rs867444811 Chr.11: 132436721-132436721 on Build GRCh38
rs560943786 Chr.11: 132436671-132436671 on Build GRCh38
rs771832798 Chr.11: 132436758-132436758 on Build GRCh38
rs539175823 Chr.11: 132436800-132436800 on Build GRCh38
rs747336860 Chr.11: 132436788-132436789 on Build GRCh38
rs778059675 Chr.11: 132436810-132436811 on Build GRCh38
rs145479085 Chr.11: 132436789-132436790 on Build GRCh38
rs755174670 Chr.11: 132436680-132436680 on Build GRCh38
rs764015633 Chr.11: 132436819-132436819 on Build GRCh38
rs373585349 Chr.11: 132436816-132436816 on Build GRCh38
rs371384830 Chr.11: 132436641-132436641 on Build GRCh38
rs138855940 Chr.11: 132436764-132436764 on Build GRCh38
rs368064190 Chr.11: 132436822-132436822 on Build GRCh38
rs369013313 Chr.11: 132436624-132436624 on Build GRCh38
rs762776894 Chr.11: 132436806-132436806 on Build GRCh38
rs377628969 Chr.11: 132436636-132436636 on Build GRCh38
rs772967983 Chr.11: 132436804-132436804 on Build GRCh38
rs775794229 Chr.11: 132436789-132436792 on Build GRCh38
rs771918593 Chr.11: 132436623-132436623 on Build GRCh38
rs771153162 Chr.11: 132436611-132436611 on Build GRCh38
rs749701062 Chr.11: 132436736-132436736 on Build GRCh38
rs753680807 Chr.11: 132436651-132436651 on Build GRCh38
rs376085210 Chr.11: 132436654-132436654 on Build GRCh38
rs762427653 Chr.11: 132436810-132436823 on Build GRCh38
rs781441114 Chr.11: 132436705-132436705 on Build GRCh38
rs987342366 Chr.11: 132436797-132436797 on Build GRCh38
rs761648096 Chr.11: 132436798-132436798 on Build GRCh38
rs890967549 Chr.11: 132436609-132436609 on Build GRCh38
rs776207806 Chr.11: 132436791-132436791 on Build GRCh38
rs769018875 Chr.11: 132436803-132436804 on Build GRCh38
rs141333534 Chr.11: 132436716-132436716 on Build GRCh38
rs759198694 Chr.11: 132436617-132436617 on Build GRCh38
rs111325555 Chr.11: 132436818-132436818 on Build GRCh38
rs995498808 Chr.11: 132436635-132436635 on Build GRCh38
rs370420226 Chr.11: 132436743-132436743 on Build GRCh38
rs764942523 Chr.11: 132436655-132436655 on Build GRCh38
rs767990932 Chr.11: 132436783-132436783 on Build GRCh38

View Full Product Details