Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45692605-45692862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QPCTL
Entrez Gene ID
54814
Gene Name
glutaminyl-peptide cyclotransferase like
Gene Aliases
gQC
UniGene
Hs.631556
Gene Chromosome Location
Chr.19: 45692402-45703990 on Build GRCh38
Transcripts
NM_001163377.1 NM_017659.3 XM_011527048.2 XM_017026900.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6760961 c.22C>A p.R8R Chr.19: 45692725-45692725 on Build GRCh38
5390108 c.37C>T p.L13L Chr.19: 45692740-45692740 on Build GRCh38
3823438 c.66C>G p.L22L Chr.19: 45692769-45692769 on Build GRCh38
4624026 c.88C>T p.L30L Chr.19: 45692791-45692791 on Build GRCh38
4079423 c.9C>G p.S3S Chr.19: 45692712-45692712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758015899 Chr.19: 45692717-45692717 on Build GRCh38
rs751330762 Chr.19: 45692757-45692757 on Build GRCh38
rs754348583 Chr.19: 45692775-45692775 on Build GRCh38
rs368538968 Chr.19: 45692787-45692787 on Build GRCh38
rs780323630 Chr.19: 45692786-45692786 on Build GRCh38
rs575048001 Chr.19: 45692717-45692718 on Build GRCh38
rs767221019 Chr.19: 45692759-45692759 on Build GRCh38
rs761765689 Chr.19: 45692682-45692682 on Build GRCh38
rs773192853 Chr.19: 45692738-45692738 on Build GRCh38
rs922122031 Chr.19: 45692732-45692732 on Build GRCh38
rs529582038 Chr.19: 45692737-45692737 on Build GRCh38
rs866401317 Chr.19: 45692813-45692813 on Build GRCh38
rs891247042 Chr.19: 45692629-45692629 on Build GRCh38
rs746112979 Chr.19: 45692730-45692730 on Build GRCh38
rs760684348 Chr.19: 45692733-45692733 on Build GRCh38
rs966971557 Chr.19: 45692678-45692678 on Build GRCh38
rs1054481075 Chr.19: 45692725-45692725 on Build GRCh38
rs766128921 Chr.19: 45692751-45692751 on Build GRCh38
rs780880329 Chr.19: 45692712-45692712 on Build GRCh38
rs770853344 Chr.19: 45692829-45692829 on Build GRCh38
rs752190389 Chr.19: 45692707-45692707 on Build GRCh38
rs560702379 Chr.19: 45692726-45692726 on Build GRCh38
rs540460807 Chr.19: 45692716-45692716 on Build GRCh38
rs762726664 Chr.19: 45692691-45692691 on Build GRCh38
rs899194856 Chr.19: 45692803-45692803 on Build GRCh38
rs772292355 Chr.19: 45692731-45692731 on Build GRCh38
rs749806467 Chr.19: 45692722-45692722 on Build GRCh38
rs990546748 Chr.19: 45692625-45692625 on Build GRCh38
rs772748704 Chr.19: 45692671-45692673 on Build GRCh38
rs544307545 Chr.19: 45692642-45692642 on Build GRCh38
rs779310706 Chr.19: 45692729-45692729 on Build GRCh38
rs533231226 Chr.19: 45692713-45692713 on Build GRCh38
rs759132086 Chr.19: 45692758-45692758 on Build GRCh38
rs1021153492 Chr.19: 45692674-45692674 on Build GRCh38
rs757747191 Chr.19: 45692777-45692777 on Build GRCh38
rs991274644 Chr.19: 45692796-45692796 on Build GRCh38
rs375377012 Chr.19: 45692785-45692785 on Build GRCh38
rs932148233 Chr.19: 45692748-45692748 on Build GRCh38
rs767415148 Chr.19: 45692840-45692840 on Build GRCh38
rs768092158 Chr.19: 45692719-45692719 on Build GRCh38
rs772089475 Chr.19: 45692679-45692679 on Build GRCh38
rs759307525 Chr.19: 45692838-45692838 on Build GRCh38
rs762927152 Chr.19: 45692747-45692747 on Build GRCh38
rs779172848 Chr.19: 45692778-45692778 on Build GRCh38
rs192761631 Chr.19: 45692686-45692686 on Build GRCh38
rs372295813 Chr.19: 45692790-45692790 on Build GRCh38
rs752257964 Chr.19: 45692764-45692764 on Build GRCh38
rs775512670 Chr.19: 45692681-45692681 on Build GRCh38
rs766185346 Chr.19: 45692704-45692704 on Build GRCh38
rs777405000 Chr.19: 45692773-45692773 on Build GRCh38
rs765105111 Chr.19: 45692684-45692684 on Build GRCh38
rs917674305 Chr.19: 45692628-45692628 on Build GRCh38
rs201142356 Chr.19: 45692788-45692788 on Build GRCh38
rs374784192 Chr.19: 45692833-45692833 on Build GRCh38
rs755750291 Chr.19: 45692765-45692765 on Build GRCh38
rs770453609 Chr.19: 45692742-45692742 on Build GRCh38

View Full Product Details