Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90644067-90644310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928909
Entrez Gene ID
101928909
Gene Name
uncharacterized LOC101928909
Gene Aliases
-
UniGene
Hs.662545
Gene Chromosome Location
Chr.14: 90642629-90648891 on Build GRCh38
Transcripts
NR_110134.1 NM_001010854.1 NM_001320421.1 XM_005267367.1 XM_011536497.1 XM_017021044.1 XM_017021045.1 XM_017021046.1 XM_017021047.1

Gene Symbol
TTC7B
Entrez Gene ID
145567
Gene Name
tetratricopeptide repeat domain 7B
Gene Aliases
TTC7L1, c14_5685
UniGene
Hs.655697
Gene Chromosome Location
Chr.14: 90540588-90816479 on Build GRCh38
Transcripts
NR_110134.1 NM_001010854.1 NM_001320421.1 XM_005267367.1 XM_011536497.1 XM_017021044.1 XM_017021045.1 XM_017021046.1 XM_017021047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1194477 c.1654C>G p.L552V Chr.14: 90644145-90644145 on Build GRCh38
6811284 c.1657C>T p.H553Y Chr.14: 90644142-90644142 on Build GRCh38
3420088 c.1717G>A p.D573N Chr.14: 90644082-90644082 on Build GRCh38
6811285 c.1657C>T p.H553Y Chr.14: 90644142-90644142 on Build GRCh38
5213476 c.1616G>A p.R539H Chr.14: 90644183-90644183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567498450 Chr.14: 90644266-90644267 on Build GRCh38
rs746947872 Chr.14: 90644253-90644254 on Build GRCh38
rs568928441 Chr.14: 90644212-90644212 on Build GRCh38
rs567119081 Chr.14: 90644276-90644281 on Build GRCh38
rs748379060 Chr.14: 90644191-90644191 on Build GRCh38
rs76773245 Chr.14: 90644257-90644257 on Build GRCh38
rs1025888139 Chr.14: 90644267-90644267 on Build GRCh38
rs750086771 Chr.14: 90644259-90644260 on Build GRCh38
rs758673151 Chr.14: 90644223-90644223 on Build GRCh38
rs775478530 Chr.14: 90644154-90644154 on Build GRCh38
rs765771489 Chr.14: 90644274-90644275 on Build GRCh38
rs111905000 Chr.14: 90644249-90644250 on Build GRCh38
rs200305180 Chr.14: 90644242-90644242 on Build GRCh38
rs762577891 Chr.14: 90644155-90644155 on Build GRCh38
rs990648052 Chr.14: 90644270-90644281 on Build GRCh38
rs372930195 Chr.14: 90644122-90644122 on Build GRCh38
rs933590166 Chr.14: 90644284-90644284 on Build GRCh38
rs754466842 Chr.14: 90644278-90644279 on Build GRCh38
rs569152988 Chr.14: 90644274-90644281 on Build GRCh38
rs1024932647 Chr.14: 90644184-90644184 on Build GRCh38
rs202158633 Chr.14: 90644245-90644245 on Build GRCh38
rs906559945 Chr.14: 90644263-90644263 on Build GRCh38
rs777433476 Chr.14: 90644258-90644261 on Build GRCh38
rs113369858 Chr.14: 90644243-90644243 on Build GRCh38
rs757729872 Chr.14: 90644087-90644087 on Build GRCh38
rs936526494 Chr.14: 90644228-90644228 on Build GRCh38
rs200054130 Chr.14: 90644095-90644095 on Build GRCh38
rs748329714 Chr.14: 90644109-90644109 on Build GRCh38
rs61739484 Chr.14: 90644098-90644098 on Build GRCh38
rs557587734 Chr.14: 90644279-90644279 on Build GRCh38
rs200928622 Chr.14: 90644244-90644244 on Build GRCh38
rs10577878 Chr.14: 90644242-90644247 on Build GRCh38
rs914743124 Chr.14: 90644271-90644271 on Build GRCh38
rs754773850 Chr.14: 90644255-90644256 on Build GRCh38
rs750822086 Chr.14: 90644166-90644166 on Build GRCh38
rs778048869 Chr.14: 90644232-90644232 on Build GRCh38
rs200679030 Chr.14: 90644259-90644259 on Build GRCh38
rs371818400 Chr.14: 90644188-90644188 on Build GRCh38
rs1053829436 Chr.14: 90644253-90644253 on Build GRCh38
rs895231441 Chr.14: 90644256-90644256 on Build GRCh38
rs376366081 Chr.14: 90644097-90644097 on Build GRCh38
rs994814512 Chr.14: 90644265-90644265 on Build GRCh38
rs147740201 Chr.14: 90644158-90644158 on Build GRCh38
rs770713403 Chr.14: 90644124-90644124 on Build GRCh38
rs71117367 Chr.14: 90644284-90644285 on Build GRCh38
rs943566359 Chr.14: 90644274-90644274 on Build GRCh38
rs573928971 Chr.14: 90644278-90644281 on Build GRCh38
rs754292263 Chr.14: 90644185-90644185 on Build GRCh38
rs781637907 Chr.14: 90644094-90644094 on Build GRCh38
rs12889670 Chr.14: 90644169-90644169 on Build GRCh38
rs759359057 Chr.14: 90644149-90644149 on Build GRCh38
rs774371261 Chr.14: 90644272-90644281 on Build GRCh38
rs371942362 Chr.14: 90644256-90644257 on Build GRCh38
rs145702221 Chr.14: 90644153-90644153 on Build GRCh38
rs902126533 Chr.14: 90644282-90644282 on Build GRCh38
rs10577879 Chr.14: 90644279-90644282 on Build GRCh38
rs3830889 Chr.14: 90644239-90644244 on Build GRCh38
rs752782457 Chr.14: 90644102-90644102 on Build GRCh38
rs769585176 Chr.14: 90644252-90644252 on Build GRCh38
rs758413326 Chr.14: 90644179-90644179 on Build GRCh38
rs776227442 Chr.14: 90644132-90644132 on Build GRCh38
rs374314608 Chr.14: 90644275-90644282 on Build GRCh38
rs549702900 Chr.14: 90644280-90644281 on Build GRCh38
rs201966954 Chr.14: 90644250-90644257 on Build GRCh38
rs974825967 Chr.14: 90644150-90644150 on Build GRCh38
rs537761782 Chr.14: 90644277-90644277 on Build GRCh38
rs796201046 Chr.14: 90644280-90644281 on Build GRCh38
rs370290759 Chr.14: 90644257-90644258 on Build GRCh38
rs1039247848 Chr.14: 90644275-90644275 on Build GRCh38
rs539444247 Chr.14: 90644283-90644283 on Build GRCh38
rs763661174 Chr.14: 90644157-90644157 on Build GRCh38
rs529112814 Chr.14: 90644163-90644163 on Build GRCh38
rs144792839 Chr.14: 90644187-90644187 on Build GRCh38
rs758340546 Chr.14: 90644210-90644211 on Build GRCh38
rs199817416 Chr.14: 90644258-90644259 on Build GRCh38
rs367676169 Chr.14: 90644183-90644183 on Build GRCh38
rs756471648 Chr.14: 90644170-90644170 on Build GRCh38
rs754299512 Chr.14: 90644277-90644278 on Build GRCh38
rs879152206 Chr.14: 90644242-90644253 on Build GRCh38
rs142561761 Chr.14: 90644164-90644164 on Build GRCh38
rs779130200 Chr.14: 90644105-90644105 on Build GRCh38
rs1286424 Chr.14: 90644281-90644281 on Build GRCh38
rs764275407 Chr.14: 90644238-90644239 on Build GRCh38

View Full Product Details